C50120
CM Sephadex® C-50
Synonym(s):
CM Sephadex®, Carboxymethyl Sephadex®
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Recommended Products
form
powder
Quality Level
technique(s)
activity assay: suitable
bead size
40-125 μm (dry)
pore size
~200,000 Da exclusion limit
operating pH
6-10
capacity
4.0-5.0 meq/g ion exchange capacity
compatibility
mode of use weak cation exchange chromatography
General description
C50120-100G′s updated product number is GE17-0220-01
Application
Sephadex C-50 is used for cation exchange media, separation media, resins, ion exchange chromatography, protein chromatography and affinity chromatography. Sephadex® C-50 has been used to study biochemical characteristics of cobra venom and effects of vitamins on lactoperoxidase enzyme activity.
Legal Information
Sephadex is a registered trademark of Cytiva
replaced by
Product No.
Description
Pricing
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
inetic studies on 1:1 electron-transfer reactions involving blue copper proteins. 15. The reactivity of <I>Anabaena variabilis</I> plastocyanin with inorganic complexes and related NMR studies.
Jackman, M.P., et al.
Journal of the American Chemical Society, 109(21), 6443-6449 (1987)
Effects of some vitamins on lactoperoxidase enzyme activity.
Sisecioglu, M., et al.
Internat. J. Vit. Nutr. Res., 79, 188-194 (2009)
Human liver acid phosphatases: purification and properties of a low-molecular-weight isoenzyme.
E M Taga et al.
Archives of biochemistry and biophysics, 214(2), 505-515 (1982-04-01)
C Chandra Sekhar et al.
Journal of biosciences, 36(2), 355-361 (2011-06-10)
A fibrinogenolytic toxin of molecular weight 6.5 kDa has been purified from the venom of Indian monocled cobra (Naja kaouthia) by repeated cation exchange chromatography on CM-sephadex C-50. The purified toxin did not show any phospholipase activity but was mildly
Purification and properties of an endoglucanase isolated from the cell walls of <I>Zea mays</I> seedlings.
Hatfield, R., and Nevis, D.J.
Carbohydrate Research, 148(2), 265-278 (1986)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service