Skip to Content
Merck
All Photos(2)

Key Documents

G25150

Sigma-Aldrich

Sephadex® G-25

Medium

Synonym(s):

Sephadex G-25 Medium, Sephadex® G-25 resins, Size exclusion resins

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
23151817
NACRES:
NA.56

Quality Level

technique(s)

buffer exchange: suitable

matrix active group

phase

swelling

1 g swells to 4-6 mL

bead size

50-150 μm

application(s)

life science and biopharma

compatibility

Cytiva

InChI

1S/C6H12O6.C3H8O3/c7-1-2-3(8)4(9)5(10)6(11)12-2;4-1-3(6)2-5/h2-11H,1H2;3-6H,1-2H2/t2-,3-,4-,5-,6-;/m1./s1

InChI key

BCPVZFFJGCURNR-OCOFDJSDSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Sephadex® G-25 Medium is a size exclusion chromatography or gel filtration resin that is created by crosslinking dextran with epichlorohydrin. The various types of Sephadex can differ in their degree of cross-linking, resulting in variances in their swelling capacity and molecular fractionation range. Sephadex G-25 is one option available among the different G-types of Sephadex. Sephadex G-25 belongs to a range of five G-types, catering to different molecule sizes. Specifically, G-10 is suitable for small molecules, while G-75 is designed to accommodate larger molecules. Sephadex G-25 provides industrial requirements by ensuring a dependable supply and offering extensive technical and regulatory assistance.

Application

Fractionation Range (MW)
Dextrans: 100 - 5,000
Globular Proteins 1,000 - 5,000

Sephadex® G-25 has been used:
  • in the desalting of protease
  • in the column for the separation of radioiodinated oTP-1 from free iodine during the characterization of radioiodinated oTP-1 and receptor assay validation
Sephadex® G-25 has been used to separate the labelled proteins from unreacted dye N-hydroxysuccinimide (NHS) esters by gel permeation chromatography.

Biochem/physiol Actions

Sephadex® G-25 is a well-established gel filtration resin, widely used for desalting and buffer exchange in industrial applications. Its hydrophilic matrix minimizes nonspecific adsorption, resulting in high recoveries during these processes for proteins and nucleic acids. Sephadex® G-25 is usually used in affinity chromatography, protein chromatography and gel filtration chromatography.

Features and Benefits

  • Rapid desalting, contaminant removal, and transfer to a new buffer in a single step.
  • High recovery rate with minimal sample dilution.
  • Offered in prepacked HiPrep Desalting and HiTrap desalting columns for convenient and fast desalting.
  • Specifically designed as a BioProcess resin for industrial applications.

Legal Information

Sephadex is a registered trademark of Cytiva

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P R Howard et al.
Clinical chemistry, 34(2), 324-330 (1988-02-01)
A monoclonal-antibody-based competitive radioimmunoassay for measuring human protein C is reported. With use of a purified protein C standard, the solid-phase assay was sensitive to less than 80 micrograms of protein C per liter. Intraassay CVs ranged from 5% to
Characterization of endometrial receptors for ovine trophoblast protein-1 during the estrous cycle and early pregnancy in sheep
Knickerbocker JJ and Niswender GD
Biology of Reproduction, 40(2), 361-369 (1989)
L R Hale et al.
Genetics, 129(1), 103-117 (1991-09-01)
Preliminary studies with restriction fragment length polymorphisms of mitochondrial DNA (mtDNA) in natural populations of Drosophila melanogaster revealed considerable variation in terms of nucleotide sequence and overall size. In this report we present data from more isofemale lines and more
K M Yocom et al.
Proceedings of the National Academy of Sciences of the United States of America, 79(22), 7052-7055 (1982-11-01)
A stable complex between pentaammineruthenium(III) and histidine-33 in horse heart ferricytochrome c is formed in the reaction between aquopentaammineruthenium(II) and the protein at pH 7. HPLC of the tryptic hydrolysate of the modified protein was employed to identify the pentaammineruthenium
Monika A Szczygiel et al.
Biology of reproduction, 67(1), 287-292 (2002-06-25)
Success with in vitro fertilization (IVF) using inbred strains of mice varies considerably and appears to be related to the proportion of motile spermatozoa present in epididymal sperm samples of different strains. In this study, motile spermatozoa were separated from

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service