Skip to Content
Merck

C3646

Sigma-Aldrich

Chitosan

from shrimp shells, ≥75% (deacetylated)

Synonym(s):

Deacetylated chitin, Poly(D-glucosamine)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.25

Pricing and availability is not currently available.

biological source

shrimp shells

Quality Level

Assay

≥75% (deacetylated)

form

powder or flakes

color

white to beige

mp

102.5  °C ((216.5 °F ))

solubility

acetic acid: water: 10 mg/mL, slightly hazy to hazy (with extensive sonication)
H2O: insoluble
organic solvents: insoluble

bulk density

0.15‑0.3 g/cm3

storage temp.

room temp

InChI

1S/C56H103N9O39/c1-87-56(86)65-28-38(84)46(19(10-74)96-55(28)104-45-18(9-73)95-49(27(64)37(45)83)97-39-12(3-67)88-47(85)20(57)31(39)77)103-54-26(63)36(82)44(17(8-72)94-54)102-53-25(62)35(81)43(16(7-71)93-53)101-52-24(61)34(80)42(15(6-70)92-52)100-51-23(60)33(79)41(14(5-69)91-51)99-50-22(59)32(78)40(13(4-68)90-50)98-48-21(58)30(76)29(75)11(2-66)89-48/h11-55,66-85H,2-10,57-64H2,1H3,(H,65,86)/t11-,12-,13-,14-,15-,16-,17-,18-,19-,20-,21-,22-,23-,24-,25-,26-,27-,28-,29-,30-,31-,32-,33-,34-,35-,36-,37-,38-,39-,40-,41-,42-,43-,44-,45-,46-,47-,48+,49+,50+,51+,52+,53+,54+,55+/m1/s1

Looking for similar products? Visit Product Comparison Guide

Application

Forms gels with multivalent anions. Gives clear solutions that dry to strong, clear films.
Shrimp shell derived chitosan, deacetylated chitin (poly β-1,4-D-glucosamine), may be used as a biocompatible, antibacterial and environmentally friendly polyelectrolyte biomaterial with a variety of applications including uses in water treatment, chromatography, additives for cosmetics, textile treatment for antimicrobial activity, novel fibers for textiles, photographic papers, biodegradable films, biomedical devices, and microcapsule implants for controlled release in drug delivery. It may be derivatized to further expand its uses.[1][2]

Features and Benefits

Biocompatible, antibacterial and environmentally friendly polyelectrolyte with a variety of applications including water treatment, chromatography, additives for cosmetics, textile treatment for antimicrobial activity, novel fibers for textiles, photographic papers, biodegradable films, biomedical devices, and microcapsule implants for controlled release in drug delivery.

Quality

May contain foreign matter.

Preparation Note

Prepared by the alkaline deacetylation of chitin obtained from the shells of shrimp (Pandalus borealis). Chitin is manufactured industrially by crushing shrimp shells, then washing the solids with acids to remove inorganic and proteinaceous material. The purified chitin is deacetylated to chitosan by treatment with a strong base such as sodium hydroxide. The resulting chitosan may be further purified by preparation of solutions in acid followed by neutralization and precipitation.

Analysis Note

The degree of acetylation may be determined by dye binding[3] or titration methods[4].

Other Notes

To gain a comprehensive understanding of our extensive range of Polysaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Need A Sample COA?

This is a sample Certificate of Analysis (COA) and may not represent a recently manufactured lot of this specific product.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sirong Tan et al.
Journal of agricultural and food chemistry, 62(8), 1866-1874 (2014-02-01)
Chitosan (CTS) and capsaicin (CAP) are two kinds of effective ingredients for antiobesity, which are extracted from crab shells and Capsicum annuum. However, the strong taste of CAP makes it difficult to consume, and the antiobesity ability of CTS is
Frederik T Larsen et al.
Vaccines, 8(2) (2020-05-21)
Infectious bronchitis virus (IBV) is a highly contagious avian coronavirus. IBV causes substantial worldwide economic losses in the poultry industry. Vaccination with live-attenuated viral vaccines, therefore, are of critical importance. Live-attenuated viral vaccines, however, exhibit the potential for reversion to
Wen-Chu Hsiao et al.
Journal of the Formosan Medical Association = Taiwan yi zhi, 118(1 Pt 1), 152-161 (2018-03-04)
Cell behaviors in three-dimensional spheroids are known to be different from those in monolayer cultures; however, very few studies have compared the characteristics of cell spheroids formed through different biomaterial-induced processes. This study investigated the mechanism of melanocyte spheroid formation
Determination of the degree of acetylation of chitin and chitosan.
Davies, D. H., and Hayes, E. R.
Methods in Enzymology, 161-pt. B, 442-446 (1988)
Sheng-Tien Li et al.
American journal of rhinology & allergy, 31(5), 289-292 (2017-09-02)
Olfactory dysfunction significantly influences patients' quality of life. Chitosan has been reported to support neuron and Schwann cell growth and even leads to orient axonal growth. However, researchers have yet to explore whether chitosan solution can promote differentiation of olfactory

Articles

Carcinogenic compounds in elastomers: 1,3-butadiene and isoprene used in elastomer manufacturing, classified as carcinogenic by IARC.

Carcinogenic compounds in elastomers: 1,3-butadiene and isoprene used in elastomer manufacturing, classified as carcinogenic by IARC.

Carcinogenic compounds in elastomers: 1,3-butadiene and isoprene used in elastomer manufacturing, classified as carcinogenic by IARC.

Carcinogenic compounds in elastomers: 1,3-butadiene and isoprene used in elastomer manufacturing, classified as carcinogenic by IARC.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service