Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
CHES
Synonym(s):
2-(Cyclohexylamino)ethanesulfonic acid
Empirical Formula (Hill Notation):
C
8
H
17
NO
3
S
CAS Number:
103-47-9
Molecular Weight:
207.29
EC Number:
203-115-6
Products
Product Category
buffers (3)
Brand
Sigma-Aldrich (3)
Feature
Formula Weight
pH Value
Physical Form
pK Value
Product Line
Purity
Quality Segment
Application
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
C2885
CHES,
≥99.0% (titration)
Expand
View Pricing
C8210
CHES,
BioXtra, ≥99.0% (titration)
Expand
View Pricing
29311
CHES,
BioUltra, ≥99.5% (T)
Expand
View Pricing