C-28009
Mesenchymal Stem Cell Growth Medium 2
Ready-to-use kit including Basal Medium and SupplementMix, 500 ml
Synonym(s):
Primary Cell Culture Media
About This Item
Recommended Products
form
liquid
packaging
pkg of 500 ml
technique(s)
cell culture | mammalian: suitable
shipped in
wet ice
1 of 4
This Item | SAB2102075 | SAB1411223 | HPA004162 |
---|---|---|---|
unconjugated | unconjugated | unconjugated | unconjugated |
rabbit | rabbit | rabbit | rabbit |
Prestige Antibodies® Powered by Atlas Antibodies | - | - | Prestige Antibodies® Powered by Atlas Antibodies |
buffered aqueous glycerol solution | buffered aqueous solution | buffered aqueous solution | buffered aqueous glycerol solution |
General description
Application
Quality
Storage and Stability
Subculture Routine
Storage Class Code
10 - Combustible liquids
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Need A Sample COA?
This is a sample Certificate of Analysis (COA) and may not represent a recently manufactured lot of this specific product.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
Are your mesenchymal stem cells really MSCs? Learn more about the MSC characterization guidelines proposed by the International Society for Cellular Therapy (ISCT).
Are your mesenchymal stem cells really MSCs? Learn more about the MSC characterization guidelines proposed by the International Society for Cellular Therapy (ISCT).
Protocols
Osteogenesis, adipogenesis and chondrogenesis differentiation protocols and media for human mesenchymal stem cell culture. Explore over 350 PromoCell products on our company website.
Osteogenesis, adipogenesis and chondrogenesis differentiation protocols and media for human mesenchymal stem cell culture. Explore over 350 PromoCell products on our company website.
Osteogenesis, adipogenesis and chondrogenesis differentiation protocols and media for human mesenchymal stem cell culture. Explore over 350 PromoCell products on our company website.
Osteogenesis, adipogenesis and chondrogenesis differentiation protocols and media for human mesenchymal stem cell culture. Explore over 350 PromoCell products on our company website.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service