Skip to Content
Merck
All Photos(1)

Key Documents

SAB1411223

Sigma-Aldrich

Anti-SALL2 antibody produced in rabbit

purified immunoglobulin, buffered aqueous solution

Synonym(s):

FLJ10414, FLJ55746, HSAL2, KIAA0360, ZNF795, p150(Sal2)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 20.9 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SALL2(6297)

General description

SALL2 is a multi-zinc finger transcription factor belonging to the Drosophila homeotic spalt-like family of developmental transcription factor genes. It is expressed during the development of human retina at the time of optic fissure closure.

Immunogen

SALL2 (ENSP00000320536, 1 a.a. ~ 198 a.a) full-length human protein.

Sequence
MAHESERSSRLGVPCGEPAELGGDASEEDHPQVCAKCCAQFTDPTEFLAHQNACSTDPPVMVIIGGQENPNNSSASSEPRPEGHNNPQVMDTEHSNPPDSGSSVPTDPTWGPERRGEESPGHFLVAATEPVCGIPVKWPAHEALEFQLHLHYHSKPGPTSAVWPRNCGWEGASNNGIQGSQGEDSPPPISASCTQGSA

Biochem/physiol Actions

SALL2 mainly associated with the growth arrest and pro-apoptotic functions. It is involved in eye morphogenesis. Alteration in the gene function causes ocular coloboma in humans and mice. It has ability to supress the c-MYC transcription by binding to the nuclease hypersensitive element of the c-MYC promoter.

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chang Kyoo Sung et al.
PloS one, 7(9), e46486-e46486 (2012-10-03)
p150, product of the SALL2 gene, is a binding partner of the polyoma virus large T antigen and a putative tumor suppressor. p150 binds to the nuclease hypersensitive element of the c-MYC promoter and represses c-MYC transcription. Overexpression of p150
Hongcang Gu et al.
Biochimica et biophysica acta, 1809(4-6), 276-283 (2011-03-03)
The product of the SALL2 protein p150(Sal2) is a multi-zinc finger transcription factor with growth arrest and proapoptotic functions that overlap those of p53. Its DNA-binding properties are unknown. We have used a modified SELEX procedure with purified p150(Sal2) and
Daniel Kelberman et al.
Human molecular genetics, 23(10), 2511-2526 (2014-01-15)
Ocular coloboma is a congenital defect resulting from failure of normal closure of the optic fissure during embryonic eye development. This birth defect causes childhood blindness worldwide, yet the genetic etiology is poorly understood. Here, we identified a novel homozygous

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service