Skip to Content
Merck
All Photos(1)

Key Documents

C2791

Sigma-Aldrich

N-(2-Chloro-4-pyridyl)-N′-phenylurea

applicable for cell culture, BioReagent

Synonym(s):

Forchlorfenuron, 4-CPPU, CPPU

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H10ClN3O
CAS Number:
Molecular Weight:
247.68
MDL number:
UNSPSC Code:
10171502
PubChem Substance ID:
NACRES:
NA.72

Quality Level

product line

BioReagent

form

powder

application(s)

agriculture

storage temp.

2-8°C

SMILES string

Clc1cc(NC(=O)Nc2ccccc2)ccn1

InChI

1S/C12H10ClN3O/c13-11-8-10(6-7-14-11)16-12(17)15-9-4-2-1-3-5-9/h1-8H,(H2,14,15,16,17)

InChI key

GPXLRLUVLMHHIK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

N-(2-Chloro-4-pyridyl)-N′-phenylurea has been used as a supplement in Murashige and Tucker medium (MT) to culture thin cell layer (TCL) explants. It has also been used to spray on to the ovaries at anthesis for induction of parthenocarpic fruit.
Forchlorfenuron (CPPU) is a cytokinin that enhances in vitro direct shoot organogenesis of Citrus aurantium L. epicotyl segments.

Biochem/physiol Actions

N-(2-Chloro-4-pyridyl)-N′-phenylurea/ CPPU, a cytokinin-like plant growth regulator participates in chlorophyll biosynthesis, cell division and cell expansion. It helps to intensify fruit set and quickens fruit enlargement. CPPU modulates carbohydrate allocation.
CPPU is a cytokinin oxidase/dehydrogenase (CKO) inhibitor.

Pictograms

Health hazardEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effect of CPPU on carbohydrate and endogenous hormone levels in young macadamia fruit
Zeng H, et al.
PLoS ONE, 11(7), e0158705-e0158705 (2016)
Masayuki Iwase et al.
Genes & genetic systems, 79(4), 199-206 (2004-10-30)
Septins, which are involved in cytokinesis, have been identified in a variety of fungi and animal cells. For analysis of the function of septin, drugs targeting septin would be useful; however, no such drugs have been available hitherto. By serendipity
Effect of 2, 4-D and 4-CPPU on somatic embryogenesis from stigma and style transverse thin cell layers of Citrus
Fiore S, et al.
Plant Cell, Tissue and Organ Culture, 68(1), 57-63 (2002)
Ying Li et al.
Journal of experimental botany, 54(385), 1245-1251 (2003-03-26)
Lagenaria leucantha is an important vegetable crop and a potential model for the study of fruit development. To study the function of D cyclins in fruit development, full-length cDNA clones for two D cyclin genes were isolated from young ovaries
Ji-Ye Hu et al.
Journal of AOAC International, 89(6), 1635-1640 (2007-01-18)
A method using solid-phase extraction for cleanup, followed by high-performance liquid chromatography with ultraviolet detection (HPLC/UV), was developed for the determination of forchlorfenuron residues in watermelon. The pesticide is extracted from the sample with acidic acetonitrile, and the extract is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service