Skip to Content
Merck
All Photos(1)

Key Documents

S7152

Sigma-Aldrich

Ser-Phe-Leu-Leu-Arg-Asn-Pro-Asn-Asp-Lys-Tyr-Glu-Pro-Phe

≥97% (HPLC), suitable for ligand binding assays and ELISA

Synonym(s):

SFLLRNPNDKYEPF, TRAP, Thrombin Receptor Activator Peptide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C81H118N20O23
CAS Number:
Molecular Weight:
1739.92
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

product name

Ser-Phe-Leu-Leu-Arg-Asn-Pro-Asn-Asp-Lys-Tyr-Glu-Pro-Phe, ≥97% (HPLC)

Quality Level

Assay

≥97% (HPLC)

form

powder

composition

Peptide content, ~70%

technique(s)

ELISA: suitable
ligand binding assay: suitable

color

white

storage temp.

−20°C

SMILES string

CC(C)CC(NC(=O)C(CC(C)C)NC(=O)C(Cc1ccccc1)NC(=O)C(N)CO)C(=O)NC(CCCNC(N)=N)C(=O)NC(CC(N)=O)C(=O)N2CCCC2C(=O)NC(CC(N)=O)C(=O)NC(CC(O)=O)C(=O)NC(CCCCN)C(=O)NC(Cc3ccc(O)cc3)C(=O)NC(CCC(O)=O)C(=O)N4CCCC4C(=O)NC(Cc5ccccc5)C(O)=O

InChI

1S/C81H118N20O23/c1-43(2)34-53(93-71(114)54(35-44(3)4)94-73(116)55(92-67(110)49(83)42-102)36-45-16-7-5-8-17-45)70(113)90-51(21-13-31-88-81(86)87)69(112)98-59(40-64(85)105)79(122)101-33-15-22-61(101)76(119)97-57(39-63(84)104)74(117)96-58(41-66(108)109)75(118)89-50(20-11-12-30-82)68(111)95-56(37-47-24-26-48(103)27-25-47)72(115)91-52(28-29-65(106)107)78(121)100-32-14-23-62(100)77(120)99-60(80(123)124)38-46-18-9-6-10-19-46/h5-10,16-19,24-27,43-44,49-62,102-103H,11-15,20-23,28-42,82-83H2,1-4H3,(H2,84,104)(H2,85,105)(H,89,118)(H,90,113)(H,91,115)(H,92,110)(H,93,114)(H,94,116)(H,95,111)(H,96,117)(H,97,119)(H,98,112)(H,99,120)(H,106,107)(H,108,109)(H,123,124)(H4,86,87,88)

InChI key

OXHYRVSBKWIFES-UHFFFAOYSA-N

Amino Acid Sequence

Ser-Phe-Leu-Leu-Arg-Asn-Pro-Asn-Asp-Lys-Tyr-Glu-Pro-Phe

Biochem/physiol Actions

Ser-Phe-Leu-Leu-Arg-Asn-Pro-Asn-Asp-Lys-Tyr-Glu-Pro-Phe has contractile effects on vascular and gastric smooth muscle. It stimulates the hydrolysis of phosphoinositides.
Thrombin receptor activator

Other Notes

Lyophilized from 0.1% TFA in H2O

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sirasate Bantuchai et al.
International journal for parasitology, 49(9), 725-735 (2019-06-28)
The malaria parasite sporozoite sequentially invades mosquito salivary glands and mammalian hepatocytes; and is the Plasmodium lifecycle infective form mediating parasite transmission by the mosquito vector. The identification of several sporozoite-specific secretory proteins involved in invasion has revealed that sporozoite
Relaxant and contractile responses of porcine pulmonary arteries to a thrombin receptor activating peptide (TRAP)
Glusa E and Manfred P
Naunyn-Schmiedeberg'S Archives of Pharmacology, 394(4), 431-443 (1994)
Xiaolin Xu et al.
Experimental animals, 68(3), 371-380 (2019-04-05)
Ketogenic diet (KD) has been used in epilepsy for decades, but previous studies found it may cause severe bone loss. Every-other-day ketogenic diet (EODKD), the combination of KD with intermittent fasting, showed better potential for seizure control recently, while its
Nobumasa Soda et al.
Journal of immunology (Baltimore, Md. : 1950), 203(5), 1356-1368 (2019-08-02)
Singleton-Merten syndrome (SMS) is a type I interferonopathy characterized by dental dysplasia, aortic calcification, skeletal abnormalities, glaucoma, and psoriasis. A missense mutation in IFIH1 encoding a cytoplasmic viral RNA sensor MDA5 has recently been identified in the SMS patients as
Sung-Hee Yoon et al.
Journal of bone and mineral research : the official journal of the American Society for Bone and Mineral Research, 34(8), 1473-1486 (2019-06-13)
The absence of functional dystrophin with mutations of the dystrophin-encoding gene in Duchenne muscular dystrophy (DMD) results in muscle inflammation and degeneration, as well as bone fragility. Long-term glucocorticoid therapy delays the muscular disease progression but suppresses growth hormone secretion

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service