Skip to Content
Merck
All Photos(1)

Key Documents

C2294

Sigma-Aldrich

Cod liver oil fatty acid methyl esters

BioReagent, suitable for insect cell culture

Synonym(s):

Fatty acid methyl esters

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352211
NACRES:
NA.75

biological source

cod liver oil

Quality Level

product line

BioReagent

form

oil

concentration

15-25% (GC)

technique(s)

cell culture | insect: suitable

impurities

≤6.0 ppt dioxin

solubility

chloroform: 100 mg/mL, clear, colorless to light yellow

shipped in

ambient

storage temp.

2-8°C

Biochem/physiol Actions

Essential fatty acids (FAs) from fish are regarded as a functional food. In particular, cod liver oil is rich in vitamin A, D, E, docosahexaenoic acid and ω3 polyunsaturated fatty acids (PUFA). Thus, cod liver oil serves as a dietary supplement. PUFA′s sourced from marine organism if helpful in the prevention of coronary heart disease. The FA composition of essential oils, is determined by its conversion to fatty acid methyl esters.

Other Notes

Natural distribution of saturated and unsaturated fatty acids as methyl esters. With 15-25% ω-3 (octadecatetraenoic, eicosapentaenoic and docosahexaenoic) methyl esters.

Preparation Note

Prepared by transmethylation of cod liver oil.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bioavailability of marine n-3 fatty acid formulations
Dyerberg J, et al.
Prostaglandins, Leukotrienes, and Essential Fatty Acids, 83(3), 137-141 (2010)
Rapid, micro-scale preparation and very fast gas chromatographic separation of cod liver oil fatty acid methyl esters
Mondello L, et al.
Journal of Pharmaceutical and Biomedical Analysis, 41(5), 1566-1570 (2006)
A gas chromatography/electron ionization- mass spectrometry- selected ion monitoring method for determining the fatty acid pattern in food after formation of fatty acid methyl esters
Thurnhofer Saskia and Vetter Walter
Journal of Agricultural and Food Chemistry, 53(23), 8896-8903 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service