Skip to Content
Merck
All Photos(2)

Key Documents

A6262

Sigma-Aldrich

O-Acetyl-L-serine hydrochloride

Synonym(s):

O-acetyl-L-serine hydrochloride (1:1)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H9NO4 · HCl
CAS Number:
Molecular Weight:
183.59
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.26

Assay

≥98.0% (TLC)

Quality Level

form

powder

technique(s)

ligand binding assay: suitable

color

white to off-white

storage temp.

−20°C

SMILES string

Cl.CC(=O)OCC(N)C(O)=O

InChI

1S/C5H9NO4.ClH/c1-3(7)10-2-4(6)5(8)9;/h4H,2,6H2,1H3,(H,8,9);1H

InChI key

MGQOSZSPKMBSRW-UHFFFAOYSA-N

Biochem/physiol Actions

O-Acetyl-L-serine, a cysteine precursor, is used as a substrate for the identification, differentiation and characterization of O-acetyl-L-serine(thiol)lyase(s) (OASTL) involved in cysteine biosynthesis.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

CysK from Lactobacillus casei encodes a protein with O-acetylserine sulfhydrylase and cysteine desulfurization activity.
Bogicevic B, Berthoud H, et al.
Applied Microbiology and Biotechnology, 94(5), 1206-1220 (2011)
Haijun Zhao et al.
The New phytologist, 211(3), 926-939 (2016-04-26)
Two low phytic acid (lpa) mutants have been developed previously with the aim to improve the nutritional value of rice (Oryza sativa) grains. In the present study, the impacts of lpa mutations on grain composition and underlying molecular mechanisms were
Agnieszka Sirko et al.
Journal of experimental botany, 55(404), 1881-1888 (2004-06-23)
The last steps of cysteine biosynthesis are catalysed by a bi-enzyme complex composed of serine acetyltransferase (SAT) and cysteine synthase, also called O-acetyl-serine (thiol) lyase (OASTL). SAT is responsible for the production of O-acetyl-serine (OAS) from serine and acetyl-coenzyme A
Anja Riemenschneider et al.
Plant physiology, 137(3), 892-900 (2005-02-25)
Plant cysteine (Cys) synthesis can occur in three cellular compartments: the chloroplast, cytoplasm, and mitochondrion. Cys formation is catalyzed by the enzyme O-acetylserine(thiol)lyase (OASTL) using O-acetylserine (OAS) and sulfide as substrates. To unravel the function of different isoforms of OASTL
Simona Carfagna et al.
Journal of plant physiology, 168(18), 2188-2195 (2011-09-17)
We analyzed the effects of deprivation and subsequent restoration of sulphate (S) in the nutrient solution on cysteine (Cys) and O-acetyl-L-serine (OAS) levels in Chlorella sorokiniana (211/8k). The removal of S from the culture medium caused a time-dependent increase in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service