Skip to Content
Merck
All Photos(1)

Key Documents

C1081

Sigma-Aldrich

8-Chloroadenosine 3′,5′-cyclic-monophosphate

≥85%

Synonym(s):

8-Chloro-cAMP, 8-Chloroadenosine 3′,5′-monophosphate, 8-Cl-cAMP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H11ClN5O6P
CAS Number:
Molecular Weight:
363.65
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.77

Assay

≥85%

form

powder

solubility

aqueous base: soluble

storage temp.

−20°C

SMILES string

Nc1ncnc2n(C3OC4COP(O)(=O)OC4C3O)c(Cl)nc12

InChI

1S/C10H11ClN5O6P/c11-10-15-4-7(12)13-2-14-8(4)16(10)9-5(17)6-3(21-9)1-20-23(18,19)22-6/h2-3,5-6,9,17H,1H2,(H,18,19)(H2,12,13,14)

InChI key

CLLFEJLEDNXZNR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Biochem/physiol Actions

Membrane-permeable cAMP analog; resistant to hydrolysis by phosphodiesterases.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vesna Vucić et al.
Investigational new drugs, 26(4), 309-317 (2007-12-07)
The antiproliferative and cytotoxic potential of the nucleotide analog 8-Cl-cAMP was tested in PC-3 and DU145 metastatic human prostate cancer cells. The drug was examined as the only therapeutic agent and in combination with ionizing irradiation (IR). Highly synergistic effects
Judith A Enyeart et al.
American journal of physiology. Endocrinology and metabolism, 301(5), E941-E954 (2011-08-04)
The regulation of cortisol synthesis and the expression of genes coding for steroidogenic proteins by 8-substituted cAMP and 8-substituted adenine derivatives were studied in bovine adrenal zona fasciculata (AZF) cells. At concentrations of 10-50 μM, 8-(4-chlorophenylthio)-cAMP (8CPT-cAMP), but not the
Audrey J Robinson-White et al.
The Journal of clinical endocrinology and metabolism, 93(3), 1020-1029 (2007-12-13)
Protein kinase A (PKA) affects cell proliferation in many cell types and is a potential target for cancer treatment. PKA activity is stimulated by cAMP and cAMP analogs. One such substance, 8-Cl-cAMP, and its metabolite 8-Cl-adenosine (8-Cl-ADO) are known inhibitors
Lela B Korićanac et al.
Annals of the New York Academy of Sciences, 1030, 384-392 (2005-01-22)
Novel antineoplastic agents, 8-chloroadenosine 3',5'-monophosphate (8-Cl-cAMP) and tiazofurin (TR), have been shown to be effective against different malignant cells. Through specific mechanisms of action they modulate the cellular signal transduction pathway, thereby causing growth inhibition, cell differentiation, and apoptosis. The
Young-Ho Ahn et al.
Cancer research, 65(11), 4896-4901 (2005-06-03)
8-Chloro-cyclic AMP (8-Cl-cAMP), which is known to induce growth inhibition, apoptosis, and differentiation in various cancer cell lines, has been studied as a putative anticancer drug. However, the mechanism of anticancer activities of 8-Cl-cAMP has not been fully understood. Previously

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service