
8-Chloroadenosine 3′,5′-cyclic-monophosphate

Synonym(s):
8-Chloro-cAMP, 8-Chloroadenosine 3′,5′-monophosphate, 8-Cl-cAMP
Empirical Formula (Hill Notation):
C10H11ClN5O6P
CAS Number:
Molecular Weight:
363.65
Products
Product Category
Brand
Formula Weight
Physical Form
Purity
Research Area
Showing 1-1 of 1
Compare
Product Number
Description
Pricing