Skip to Content
Merck
All Photos(2)

Key Documents

B8895

Sigma-Aldrich

Anti-Rabbit IgG (whole molecule)–Biotin antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

goat

Quality Level

conjugate

biotin conjugate

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous solution

technique(s)

direct ELISA: 1:200,000
immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:800
western blot: 1:400,000-1:800,000 using using an assay detecting actin in total cell extract of HeLa cells (5-10 μg per well)

storage temp.

−20°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

IgG antibodies regulate several functions such as phagocytosis and complement activation. Thus they play a crucial role in facilitating cytological immune responses. Anti-rabbit IgG (whole molecule)–biotin antibody can be used in immunohistochemistry. The product shows minimal interspecies cross reactivity to human IgG hence can be used in enzyme immunoassays or dot blot in presence of human IgG. Anti-rabbit IgG (whole molecule)–biotin antibody reacts specifically to all rabbit immunoglobulin but has no reactivity with human IgG.
IgGs are glycoprotein antibodies that modulate several immune responses. Rabbit IgGs against target proteins are often used as primary antibodies in various research applications. Thus, secondary anti-rabbit IgGs conjugated to a detectable substrate are useful tools for the analysis of target proteins
Anti-Rabbit IgG (whole molecule)-Biotin antibody reacts with rabbit serum and rabbit IgG. The product does not cross-react with human IgG.

Specificity

Binds all rabbit Igs.

Immunogen

Purified rabbit IgG

Application

Anti-Rabbit IgG (whole molecule)-Biotin antibody has been used in
  • Immunofluorescence
  • Immunohistochemistry
  • Western blot
  • Direct enzyme linked immunosorbent assay (ELISA)

Biochem/physiol Actions

Avidin, Streptavidin or ExtrAvidine show strong affinity interaction with biotin rendering the biotin-avidin system an extremely effective tool in molecular biology, protein chemistry and immunology. The high specificity of the biotinylated antibody to rabbit IgG and the stability of the biotin-avidin complex has been exploited in variety of reagents (Avidin, Streptavidin or ExtrAvidin conjugated to FITC, TRITC, Peroxidase or Alkaline Phosphatase), for the detection and quantitation of rabbit IgG.

Other Notes

Antibody adsorbed with human IgG.

Physical form

Solution in 0.01 M phosphate buffered saline pH 7.4, containing 15 mM sodium azide

Preparation Note

Adsorbed to reduce background staining with human samples.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jeffery M Vahrenkamp et al.
Cell reports, 22(11), 2995-3005 (2018-03-15)
Steroid hormone receptors are simultaneously active in many tissues and are capable of altering each other's function. Estrogen receptor α (ER) and glucocorticoid receptor (GR) are expressed in the uterus, and their ligands have opposing effects on uterine growth. In
Leonurine prevents atherosclerosis via promoting the expression of ABCA1 and ABCG1 in a Ppargamma/Lxralpha signaling pathway-dependent manner
<BIG>Jiang T, et al.</BIG>
Cellular Physiology and Biochemistry, 43, 1703-1717 (2017)
Feng Yang et al.
Cancer research, 65(19), 8887-8895 (2005-10-06)
Our previous studies have defined reactive stroma in human prostate cancer and have developed the differential reactive stroma (DRS) xenograft model to evaluate mechanisms of how reactive stroma promotes carcinoma tumorigenesis. Analysis of several normal human prostate stromal cell lines
Jenni Leppiniemi et al.
PloS one, 6(1), e16576-e16576 (2011-02-10)
The extensive use of avidin and streptavidin in life sciences originates from the extraordinary tight biotin-binding affinity of these tetrameric proteins. Numerous studies have been performed to modify the biotin-binding affinity of (strept)avidin to improve the existing applications. Even so
Thomas H Priddle et al.
Cerebral cortex (New York, N.Y. : 1991), 23(8), 1933-1941 (2012-06-30)
Protocadherins 11X and 11Y are cell adhesion molecules of the δ1-protocadherin family. Pcdh11X is present throughout the mammalian radiation; however, 6 million years ago (MYA), a reduplicative translocation of the Xq21.3 block onto what is now human Yp11 created the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service