AXYT200YRS
Axygen® Universal Fit Pipet Tip
racked, size 200 μL, yellow, pkg of 5x960tips/cs, sterile
Synonym(s):
Pipette tips, universal fit, Pipette tips, pipette tips
About This Item
Recommended Products
material
yellow
sterility
sterile
feature
DNA free
RNase free
barrier: no
endotoxin free
filter: no
manufacturer/tradename
Axygen Inc T-200-Y-R-S
Corning
packaging
pkg of 5x960tips/cs
size
200 μL
color
yellow
Looking for similar products? Visit Product Comparison Guide
1 of 4
This Item | AXYT1000B | AXYT200CL | AXYT200Y |
---|---|---|---|
sterile | non-sterile | non-sterile | non-sterile |
Corning, Axygen Inc T-200-Y-R-S | Axygen Inc T-1000-B, Corning | Axygen Inc T-200-C-L, Corning | Axygen Inc T-200-Y, Corning |
yellow | blue | clear | yellow |
pkg of 5x960tips/cs | pkg of 5x1000tips/cs | pkg of 20x1000tips/cs | pkg of 20x1000tips/cs |
200 μL | - | 200 μL | 200 μL |
Features and Benefits
- Beveled orifice for easy delivery
- Error-free graduation marks
- Applicable for sensitive biological experiments
Other Notes
Legal Information
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service