Skip to Content
Merck
All Photos(1)

Key Documents

CLS3599

Corning® 96 Well TC-Treated Microplates

size 96 wells, polystyrene, flat bottom, case of 100 (individually wrapped), sterile, lid

Synonym(s):

96 well, cell culture plate, multiple well plates, multiwell plates, not treated, tissue culture plates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122107
eCl@ss:
32190102
NACRES:
NB.13

material

clear bottom
flat bottom
polystyrene
round clear wells

description

growth area .32 cm2

sterility

sterile; γ-irradiated by SER-TAIN process
sterile

feature

lid
skirt

packaging

case of 100 (individually wrapped)

manufacturer/tradename

Corning 3599

size

96 wells

well diam.

6.4 mm

well volume

360 μL

well working volume

75-200 μL

color

clear

binding type

Tissue Culture (TC)-treated surface

Looking for similar products? Visit Product Comparison Guide

General description

Rings on lid prevent cross-contamination. All plates have a uniform footprint and a raised bead to aid stacking. Alphanumerics provide well identification. Polystyrene unless stated otherwise.

Application

Corning® 96 Well TC-Treated Microplates have been used for:
  • dried blood spot (DBS) assay protocol for eluting DBS samples and DBS controls overnight
  • in vitro cell growth and invasion assays
  • ex vivo cell culture

Features and Benefits

  • Enhanced cell attachment:Specifically treated surface to promote optimal cell adhesion and growth.
  • Reliablesterilization: Gamma radiation sterilized and guaranteed nonpyrogenic for safecell culture applications.
  • Convenientstacking: Uniform footprint and raised bead design for easy and secure platestacking.
  • Hygienic packaging: Individually wrapped foradded protection and cleanliness.

Legal Information

Corning is a registered trademark of Corning, Inc.
SER-TAIN is a trademark of Sigma-Aldrich Co. LLC

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amelia E Sancilio et al.
Scientific reports, 11(1), 15321-15321 (2021-07-30)
The spike protein of SARS-CoV-2 engages the human angiotensin-converting enzyme 2 (ACE2) receptor to enter host cells, and neutralizing antibodies are effective at blocking this interaction to prevent infection. Widespread application of this important marker of protective immunity is limited
Mark Marsland et al.
International journal of molecular sciences, 24(2) (2023-01-22)
Glioblastoma multiforme (GBM) is the most lethal adult brain cancer. Temozolomide (TMZ), the standard chemotherapeutic drug used in GBM, has limited benefit and alternate therapies are needed to improve GBM treatment. Nerve growth factor (NGF) and its precursor proNGF are
Competitive inhibition of IL-2/IL-2R has a dual effect on HSC ex vivo expansion and IL-2R (CD25) content
Merve Uslu, et al.
International Immunopharmacology (2022)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service