AXYSCT10MLS
Screw-cap tubes and caps, self-standing, unassembled
capacity 10 mL, blue, sterile
Synonym(s):
centrifuge tubes
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121800
NACRES:
NB.22
Recommended Products
material
blue
conical bottom tube (self-standing)
screw top cap
sterility
sterile
packaging
pack of 25 ea
case of 500 ea
manufacturer/tradename
Axygen Inc SCT-10ML-S
Corning
capacity
10 mL
color
clear
fitting
cap (screw cap)
Looking for similar products? Visit Product Comparison Guide
Related Categories
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service