Skip to Content
Merck
All Photos(2)

Key Documents

AV50523

Sigma-Aldrich

Anti-SPRY2 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-MGC23039, Anti-Sprouty homolog 2 (Drosophila), Anti-hSPRY2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

35 kDa

species reactivity

rat, dog, pig, mouse, human

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SPRY2(10253)

General description

The gene SPRY2 (Sprouty homolog 2) is mapped to human chromosome 13q31.1. SPRY2 localizes in the cytoplasm and co-localizes with microtubules.

Immunogen

Synthetic peptide directed towards the N terminal region of human SPRY2

Application

Anti-SPRY2 antibody produced in rabbit is suitable for western blotting at a concentration of 1.0μg/ml.

Biochem/physiol Actions

Sprouty proteins are inhibitors of receptor tyrosine kinase signaling proteins and mediators of organogenesis, branching of lung alveoli and endothelial tubes. They inhibit the activity of Ras/Erk pathway. SPRY2 (Sprouty homolog 2) is bimodal regulator of epidermal growth factor receptor (EGFR)/mitogen-activated protein kinase signaling. SPRY2 is down-regulated in epithelial ovarian cancer. Down-regulation of SPRY2 in high-grade serous ovarian carcinoma promotes EGF-induced E-cadherin levels, thereby resulting in cell invasion.

Sequence

Synthetic peptide located within the following region: MEARAQSGNGSQPLLQTPRDGGRQRGEPDPRDALTQQVHVLSLDQIRAIR

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Samar Masoumi-Moghaddam et al.
International journal of cancer, 137(3), 560-570 (2015-01-30)
Sprouty proteins are evolutionary-conserved modulators of receptor tyrosine kinase signaling, deregulation of which has been implicated in the pathophysiology of cancer. In the present study, the expression status of Spry2 and Spry4 proteins and its clinical relevance in human epithelial
Wai-Kin So et al.
FEBS letters, 589(3), 302-309 (2014-12-24)
Sprouty (SPRY) proteins are well-characterized factors that inhibit receptor tyrosine kinase signaling. Our Human Exonic Evidence-Based Oligonucleotide (HEEBO) microarray results showed that the mRNA levels of SPRY2, but not of SPRY1 or SPRY4, are down-regulated in high-grade serous ovarian carcinoma
Chloé Quélin et al.
European journal of medical genetics, 52(1), 41-46 (2008-11-22)
13q deletion is characterized by a wide phenotypic spectrum resulting from a partial deletion of the long arm of chromosome 13. The main clinical features are mental retardation, growth retardation, craniofacial dysmorphy and various congenital defects. Only one recent Italian
J Lim et al.
The Journal of biological chemistry, 275(42), 32837-32845 (2000-07-11)
Sprouty (Spry) was first identified in a genetic screen in Drosophila to be an antagonist of fibroblast growth factor and epidermal growth factor (EGF) signaling, seemingly by inhibiting the Ras/MAP kinase pathway. Data base searches lead to the identification and
G R Guy et al.
The Journal of endocrinology, 203(2), 191-202 (2009-05-09)
Sprouty proteins are involved in organogenesis, particularly during the branching of endothelial tubes, and existing evidence suggests that Sprouty's point of action lies downstream of receptor signaling to inhibit the activation of the central Ras/Erk pathway. How Sprouty proteins accomplish

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service