Skip to Content
Merck
All Photos(1)

Key Documents

A9519

Sigma-Aldrich

N-Acetylmuramyl-L-alanyl-D-isoglutamine hydrate

≥98% (TLC), suitable for ligand binding assays

Synonym(s):

Adjuvant Peptide, Muramyl dipeptide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H32N4O11 · xH2O
CAS Number:
Molecular Weight:
492.48 (anhydrous basis)
Beilstein:
4220745
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

product name

N-Acetylmuramyl-L-alanyl-D-isoglutamine hydrate, ≥98% (TLC)

Quality Level

Assay

≥98% (TLC)

form

powder

technique(s)

ligand binding assay: suitable

color

white

application(s)

cell analysis

storage temp.

−20°C

SMILES string

C[C@H](NC(=O)[C@@H](C)O[C@H]1[C@H](O)[C@@H](CO)OC(O)[C@@H]1NC(C)=O)C(=O)N[C@H](CCC(O)=O)C(N)=O

InChI

1S/C19H32N4O11/c1-7(17(30)23-10(16(20)29)4-5-12(26)27)21-18(31)8(2)33-15-13(22-9(3)25)19(32)34-11(6-24)14(15)28/h7-8,10-11,13-15,19,24,28,32H,4-6H2,1-3H3,(H2,20,29)(H,21,31)(H,22,25)(H,23,30)(H,26,27)/t7-,8+,10+,11+,13+,14+,15+,19?/m0/s1

InChI key

BSOQXXWZTUDTEL-QAQREVAFSA-N

Looking for similar products? Visit Product Comparison Guide

Amino Acid Sequence

MurNAc-Ala-Agn

Application

N-Acetylmuramyl-L-alanyl-D-isoglutamine hydrate has been used in staphylococcal cell wall preparation to induce the release of TNF from human monocytes.

Biochem/physiol Actions

Enhances antigenicity of weak antigens, increasing antibody titer.
Muramyl peptides are monomeric components that make up the bacterial cell wall peptidoglycan. It is generally produced by mammalian macrophages, as an immune response in the breakdown of bacterial cell wall. MDP promotes the production of interleukin-1 and tumor necrosis factor (TNF), which increase slow-wave sleep.
N-Acetylmuramyl-L-alanyl-D-isoglutamine (MDP, muramyl dipeptide), a neurotrophic and immunomodulatory factor related to bacterial peptidoglycans, is used as an immune modulation factor that enhances immune response and to study neural development.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shoichi Kusumoto et al.
Proceedings of the Japan Academy. Series B, Physical and biological sciences, 86(4), 322-337 (2010-05-01)
Chemistry-based investigation is reviewed which led to identification of the active entities responsible for the immunostimulating potencies of peptidoglycan and lipopolysaccharide. Though these glycoconjugates which ubiquitously occur in wide range of bacteria as the essential components of their cell envelopes
Muramyl dipeptide and IL-1 effects on sleep and brain temperature after inhibition of serotonin synthesis.
Imeri L
The American Journal of Physiology, 273(5 Pt 2), R1663-R1668 (1997)
Jesse M Hansen et al.
The Journal of infectious diseases, 209(7), 1045-1054 (2013-11-23)
Mycobacteria produce an unusual, glycolylated form of muramyl dipeptide (MDP) that is more potent and efficacious at inducing NOD2-mediated host responses. We tested the importance of this modified form of MDP in Mycobacterium tuberculosis by disrupting the gene, namH, responsible
Hui-Qing Liu et al.
Arteriosclerosis, thrombosis, and vascular biology, 33(9), 2193-2201 (2013-07-23)
The activity of eicosanoid pathways is critical to the inflammatory and immune responses that are associated with the progression of atherosclerosis. Yet, the signals that regulate these pathways are poorly understood. Here, we address whether the innate immune signals of
Mohammad Salem et al.
Cellular and molecular life sciences : CMLS, 70(18), 3391-3404 (2013-01-01)
Crohn's disease (CD) is one of main disease entities under the umbrella term chronic inflammatory bowel disease. The etiology of CD involves alterations in genetic, microbiological, and immunological factors. This review is devoted to the role of the bacterial wall

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service