Skip to Content
Merck
All Photos(2)

Key Documents

A9348

ART® self-sealing barrier pipette tips

ART 1000G, volume range 100-1000 μL, sterile

Synonym(s):

pipet tips, pipette tips

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
NACRES:
NB.01
volume:
100—1000 μL
sterility:
sterile
feature:
DNase free
RNase free
barrier
packaging:
case of 4 packs
pack of 800 ea
manufacturer/tradename:
Molecular BioProducts 2079G

material

colorless

sterility

sterile

feature

DNase free
RNase free
barrier

packaging

case of 4 packs
pack of 800 ea

manufacturer/tradename

Molecular BioProducts 2079G

technique(s)

PCR: suitable

maximum volume

1 mL

volume range

100-1000 μL

volume

100—1000 μL

compatibility

for use with

  • Biohit Variable 200-1000 μl
  • Biohit Proline-Electronic 1000 μl
  • Capp Aero 1000 μl
  • Corning 4964 100-1000 μl
  • Eppendorf 4810 100-1000 μl
  • Eppendorf Research/Reference 100-1000 μl
  • Eppendorf Response 4850 20-500 μl
  • Finnpipette Digital 4027/45 200-1000 μl
  • Mettler-Toledo VoluMate 100-1000 μl
  • Nichiryo BenchMate 100-1000 μl
  • Pipetman P-1000 100-1000 μl
  • Rainin EDP-Series 1000 100-1000 μl
  • Socorex/Wheaton Calibra/822 100-1000 μl
  • Transferpette 100-1000 μl

Looking for similar products? Visit Product Comparison Guide

General description

ART® self-sealing barrier pipette tips can be used to achieve precise and consistent performance in your applications. These tips create a reliable seal when exposed to potential contaminants, effectively trapping them inside the barrier for enhanced protection. Designed to assist in pipetting across various applications, ART tips offer versatility and reliable results.

Application

ART® self-sealing barrier pipette tips have been used in PCR reactions and LC–MS/MS (liquid chromatography–mass spectrometry).

Features and Benefits

  • Sterile tips are guaranteed to be free from DNase, RNase, DNA, ATP, PCR inhibitors, and endotoxins.
  • Available in sterile and non-sterile options to accommodate different requirements.
  • Offered in a variety of packaging options for convenience and flexibility.
  • Manufactured in accordance with the industry′s highest quality standards.
  • Wide orifice genomic tips designed specifically for handling genomic DNA, fragile cell lines, and other viscous materials.

Packaging

960EA = 10 trays of 96 tips (1 pack)
4800EA = 5 packs of 960 tips (50 trays of 96 tips)

800EA = 8 trays of 100 tips (1pack)
3200EA = 4 packs of 800EA

Other Notes

Browse all pipette tips

Legal Information

ART is a registered trademark of Molecular BioProducts, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nitrogenase genes in PCR and RT-PCR reagents: implications for studies of diversity of functional genes.
Zehr JP
Biotechniques, 35, 996-1002 (2003)
Prolonged arthritis associated with sindbis-related (Pogosta) virus infection.
Laine M
Rheumatology (Oxford, England), 39, 1272-1274 (2000)
Quantitative profiling of endocannabinoids in lipoproteins by LC-MS/MS.
Bilgin M, et al.
Analytical and Bioanalytical Chemistry, 407, 5125-5131 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service