Skip to Content
Merck
All Photos(1)

Key Documents

Z717193

ART® self-sealing barrier pipette tips

ART 1000 REACH, maximum volume 1000 μL, sterile

Synonym(s):

pipet tips, pipette tips

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121601
NACRES:
NB.01
maximum volume:
1000 μL
sterility:
sterile
feature:
DNase free
RNase free
barrier
packaging:
pack of 800 ea
manufacturer/tradename:
Molecular BioProducts 2079
Pricing and availability is not currently available.

material

colorless

sterility

sterile

feature

DNase free
RNase free
barrier

packaging

pack of 800 ea

manufacturer/tradename

Molecular BioProducts 2079

technique(s)

PCR: suitable

maximum volume

1000 μL

Looking for similar products? Visit Product Comparison Guide

Packaging

960EA = 10 trays of 96 tips (1 pack)
4800EA = 5 packs of 960 tips (50 trays of 96 tips)

800EA = 8 trays of 100 tips (1pack)
3200EA = 4 packs of 800EA

Compatibility

Compatibility
Biohit mLINE 100-1000 μl
BioHit Proline Variable
Eppendorf Research/Reference
Gilson/Pipetman P-1000
Labsystems 4027/4500
Nichiryo/Oxford Benchmate
Socorex Calibra

Other Notes

Browse all pipette tips

Legal Information

ART is a registered trademark of Molecular BioProducts, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service