Skip to Content
Merck
All Photos(1)

Key Documents

A8861

Sigma-Aldrich

Aceclofenac

≥98% (HPLC)

Synonym(s):

2-[(2,6-Dichlorophenyl)amino]benzeneacetic acid carboxymethyl ester;

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H13Cl2NO4
CAS Number:
Molecular Weight:
354.18
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

off-white to light tan

solubility

DMSO: ≥20 mg/mL

storage temp.

room temp

SMILES string

OC(=O)COC(=O)Cc1ccccc1Nc2c(Cl)cccc2Cl

InChI

1S/C16H13Cl2NO4/c17-11-5-3-6-12(18)16(11)19-13-7-2-1-4-10(13)8-15(22)23-9-14(20)21/h1-7,19H,8-9H2,(H,20,21)

InChI key

MNIPYSSQXLZQLJ-UHFFFAOYSA-N

Gene Information

human ... PTGS2(5743)

Looking for similar products? Visit Product Comparison Guide

Application

Aceclofenac (ACE) has been used as an internal standard in the in vivo blood-brain barrier assay. It has also been used to study the interaction of ACE with bovine serum albumin (BSA) by using spectroscopic techniques in combination with computational methods.

Biochem/physiol Actions

Aceclofenac is a phenyl acetic acid derivative used for treating symptoms like swelling, tenderness, and stiffness due to muscle-skeletal and bone-related diseases (rheumatoid arthritis, juvenile arthritis, osteoarthritis, and acute gouty arthritis). This compound shows analgesic and antipyretic properties. Aceclofenac suppresses prostaglandin biosynthesis.
Non-steroidal, anti-inflammatory drug (NSAID), with selectivity for COX-2 over COX-1.

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

An insight into the binding of aceclofenac with bovine serum albumin at physiological condition: a spectroscopic and computational approach.
Neeraj Dohare et al.
Journal of biomolecular structure & dynamics, 36(2), 398-406 (2017-01-25)
Viney Chawla et al.
Colloids and surfaces. B, Biointerfaces, 92, 293-298 (2012-01-10)
Solid lipid nanoparticles (SLN) of aceclofenac were prepared using Taguchi experimental design by Trotta method. The prepared SLN were formulated into a gel preparation, using carbopol 940. Gels were evaluated for drug content, bioadhesion and their stability against change of
M Dooley et al.
Drugs, 61(9), 1351-1378 (2001-08-21)
Aceclofenac is an orally administered phenylacetic acid derivative with effects on a variety of inflammatory mediators. Through its analgesic and anti-inflammatory properties, aceclofenac provides symptomatic relief in a variety of painful conditions. In patients with osteoarthritis of the knee, the
Anil Pareek et al.
The journal of pain : official journal of the American Pain Society, 12(5), 546-553 (2011-02-01)
The efficacy and safety of aceclofenac control release (CR) tablets was compared with conventional aceclofenac tablets in patients with knee osteoarthritis (OA). This was a double-blind, double-dummy, randomized, parallel group multicentric study conducted at 6 centers. Two hundred and eighty
Sandipan Dasgupta et al.
Expert opinion on drug delivery, 10(4), 411-420 (2013-01-16)
The aim of the present study was to investigate the potential of a nanoemulsion for topical delivery of aceclofenac using different excipients having optimum emulsifying ability rather than their solubilizing capacity. The oil-in-water nanoemulsions were prepared by screening the excipients

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service