Skip to Content
Merck
All Photos(2)

Key Documents

93484

Supelco

Diclofenac sodium salt

analytical standard

Synonym(s):

2-[(2,6-Dichlorophenyl)amino]benzeneacetic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H10Cl2NNaO2
CAS Number:
Molecular Weight:
318.13
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.5% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤2.0% water

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

SMILES string

[Na+].[O-]C(=O)Cc1ccccc1Nc2c(Cl)cccc2Cl

InChI

1S/C14H11Cl2NO2.Na/c15-10-5-3-6-11(16)14(10)17-12-7-2-1-4-9(12)8-13(18)19;/h1-7,17H,8H2,(H,18,19);/q;+1/p-1

InChI key

KPHWPUGNDIVLNH-UHFFFAOYSA-M

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Standard NSAID and cyclooxygenase (COX) inhibitor. Major metabolites are 4´-hydroxydiclofenac and 5´-hydroxydiclofenac. Has been used as substrate selective for CYP2C9.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 2 - Repr. 2 - STOT RE 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jorgelindo da Veiga Moreira et al.
Scientific reports, 9(1), 3153-3153 (2019-03-01)
In the recent years, cancer research succeeded with sensitive detection methods, targeted drug delivery systems, and the identification of a large set of genes differently expressed. However, although most therapies are still based on antimitotic agents, which are causing wide
D Aygün et al.
Histology and histopathology, 27(4), 417-436 (2012-03-01)
Nonsteroidal anti-inflammatory drugs (NSAIDs) are widely used for the purpose of anti-inflammation, antipyretic, and analgesia. For this aim, they are used for the alleviation of pain, fever, and inflammation associated with rheumatoid arthritis, sports injuries, and temporary pain. However, treatment
R N Brogden et al.
Drugs, 20(1), 24-48 (1980-07-01)
Diclofenac sodium, a phenylacetic acid derivative, is a non-steroidal, anti-inflammatory, analgesic agent advocated for use in rheumatoid arthritis, degenerative joint disease, ankylosing spondylitis and allied conditions, and in the treatment of pain resulting from minor surgery, trauma and dysmenorrhoea. Published
P A Todd et al.
Drugs, 35(3), 244-285 (1988-03-01)
Diclofenac is a non-steroidal anti-inflammatory drug (NSAID) advocated for use in painful and inflammatory rheumatic and certain non-rheumatic conditions. It is available in a number of administration forms which can be given orally, rectally or intramuscularly. Conveniently, dosage adjustments are
Paul L McCormack et al.
Drugs, 68(1), 123-130 (2007-12-18)
*A new formulation of the nonselective NSAID diclofenac sodium suitable for intravenous bolus injection has been developed using hydroxypropyl beta-cyclodextrin as a solubility enhancer (HPbetaCD diclofenac). * HPbetaCD diclofenac intravenous bolus injection was shown to be bioequivalent to the existing

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service