Skip to Content
Merck
All Photos(1)

Key Documents

A8264

Sigma-Aldrich

4-Aminophthalhydrazide

≥98% (HPLC), powder

Synonym(s):

6-Amino-2,3-dihydro-1,4-phthalazinedione, Isoluminol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H7N3O2
CAS Number:
Molecular Weight:
177.16
Beilstein:
164804
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.83

Product Name

4-Aminophthalhydrazide,

Assay

≥98% (HPLC)

Quality Level

form

powder

mp

300 °C (lit.)

solubility

2 M NH4OH: 50 mg/mL, clear to slightly hazy

SMILES string

Nc1ccc2C(=O)NNC(=O)c2c1

InChI

1S/C8H7N3O2/c9-4-1-2-5-6(3-4)8(13)11-10-7(5)12/h1-3H,9H2,(H,10,12)(H,11,13)

InChI key

HUDPLKWXRLNSPC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

4-Aminophthalhydrazide has been used in:
  • chemiluminescence-based reactive oxygen species (ROS) assay in bone marrow (BM) neutrophils post-Yersinia pseudotuberculosis infection
  • neutrophil ROS assays post-K. pneumonia exposure
  • neutrophil ROS assays in response to formylated-methionine-leucine-proline

Biochem/physiol Actions

4-Aminophthalhydrazide or isoluminol is a chemiluminescent compound that comprises a phthalazine ring. It is more hydrophilic than luminol. Isoluminol is commercially used in a wide variety of in vitro immunoassays and is a chemiluminescence amplifier for studying reactive oxygen species. It is also useful as an electroluminescence (ECL) biosensor and as a probe for rapid microbial detection.

Features and Benefits

Produces chemiluminescence.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Viera Jancinová et al.
Redox report : communications in free radical research, 11(3), 110-116 (2006-06-30)
To address the question why isoluminol, but not luminol, failed to detect oxidants produced intracellularly, differences between these luminophores were investigated with respect to physicochemical parameters and the character of chemiluminescence signal. Our results showed the isoluminol molecule to be
Caifeng Ding et al.
Chemical communications (Cambridge, England), 46(42), 7990-7992 (2010-09-25)
A new strategy of photoelectrochemical (PEC) analysis with chemiluminescence (CL) of the isoluminol-H(2)O(2)-Co(2+) system as a light source for the determination of physiological thiols in cancer cells was reported for the first time.
B G Yushkov et al.
Bulletin of experimental biology and medicine, 150(3), 352-354 (2011-01-18)
The effects of 3-aminophthalhydrazide and carrageenan on reparative regeneration and expression of CD117 by liver cells after partial hepatectomy were studied in mice. 3-Aminophthalhydrazide stimulated regeneration of the liver and increased the count of CD117(+) hepatocytes. By contrast, carrageenan inhibited
Kazuhiko Tsukagoshi et al.
Journal of chromatography. A, 1143(1-2), 288-290 (2007-02-03)
In our previous study, we developed capillary electrophoresis with an ultraviolet absorption/chemiluminescence dual detection system. Here, to demonstrate one of the possible applications of the capable system as well as confirm the advantage, migration behavior of isoluminol isothiocyanate-labeled alpha-amino acids
M T Abidov et al.
Bulletin of experimental biology and medicine, 153(1), 61-63 (2012-07-19)
The content of CD117(+)cells in the kidneys and CD45(low)CD117(+)cells in the bone marrow and blood of mice were studied after partial nephrectomy and under conditions of macrophage stimulation with 3-aminophthalhydrazide. The counts of tubular CD117(+)epitheliocytes sharply increased and the content

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service