Skip to Content
Merck
All Photos(1)

Key Documents

25455

Sigma-Aldrich

4-Chloro-7-nitrobenzofurazan

BioReagent, suitable for fluorescence, ≥97.0% (HPLC)

Synonym(s):

4-Chloro-7-nitro-1,2,3-benzoxadiazole, NBD-chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H2ClN3O3
CAS Number:
Molecular Weight:
199.55
Beilstein:
614212
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.32
Pricing and availability is not currently available.

product line

BioReagent

Quality Level

Assay

≥97.0% (HPLC)

mp

97-100 °C
97-99 °C (lit.)

fluorescence

λex 336 nm in methanol
λex 420 nm; λem ~540 nm in ethanol (after derivatization with glycine)

suitability

suitable for fluorescence

SMILES string

[O-][N+](=O)c1ccc(Cl)c2nonc12

InChI

1S/C6H2ClN3O3/c7-3-1-2-4(10(11)12)6-5(3)8-13-9-6/h1-2H

InChI key

IGHBXJSNZCFXNK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

4-Chloro-7-nitrobenzofurazan (NBD-chloride) is a fluorescence assay reagent used to label free sulfhydryls and N-terminals within proteins. NBD-chloride is used as an enzyme inhibitor and as a trapping agent for cysteine sulfenic acid.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Leslie B Poole et al.
Methods in enzymology, 348, 122-136 (2002-03-12)
C165S AhpC in its sulfenate (Cys-SO-) and presumed thiolate (Cys-S-) forms at pH 7 (pKa for sulfenic acid about pH 6.1) exhibit low extinction absorbance bands around 367 and 324 nm, respectively. Sulfenic acid content of the protein can be
P D Bragg et al.
Biochimica et biophysica acta, 1413(3), 159-171 (1999-11-11)
Pyridine nucleotide transhydrogenases of bacterial cytosolic membranes and mitochondrial inner membranes are proton pumps in which hydride transfer between NADP(+) and NAD(+) is coupled to proton translocation across cytosolic or mitochondrial membranes. The pyridine nucleotide transhydrogenase of Escherichia coli is
Rob van Zwieten et al.
Biochimica et biophysica acta, 1821(12), 1493-1500 (2012-09-11)
Maintenance of the asymmetric distribution of phospholipids across the plasma membrane is a prerequisite for the survival of erythrocytes. Various stimuli have been shown to induce scrambling of phospholipids and thereby exposure of phosphatidylserine (PS). In two types of patients
Elham Raeisi et al.
The Journal of membrane biology, 245(10), 633-642 (2012-08-14)
This study investigated whether molecules spontaneously transported inside cells, like glucose derivatives, can also be used as electropermeabilization markers. Uptake of a fluorescent deoxyglucose derivative (2-NBDG) by normal and electropermeabilized cells in culture was analyzed. 2-NBDG was added to DC-3F
Rolffy Ortiz-Andrade et al.
Journal of ethnopharmacology, 143(2), 455-462 (2012-07-24)
Justicia spicigera is a plant species used for the Teenak (Huesteca Potosina) and Mayan (Yucatan peninsula) indigenous for the empirical treatment of diabetes, infections and as stimulant. To evaluate the cytotoxicity, antioxidant and antidiabetic properties of J. spicigera. The effects

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service