Skip to Content
Merck
All Photos(1)

Key Documents

A7229

Sigma-Aldrich

PUGNAc

≥95% (HPLC)

Synonym(s):

O-(2-Acetamido-2-deoxy-D-glucopyranosylidenamino) N-phenylcarbamate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H19N3O7
CAS Number:
Molecular Weight:
353.33
Beilstein:
4274031
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.26

Quality Level

Assay

≥95% (HPLC)

form

crystals

storage temp.

−20°C

SMILES string

CC(=O)N[C@@H]1[C@@H](O)[C@H](O)[C@@H](CO)O\C1=N/OC(=O)Nc2ccccc2

InChI

1S/C15H19N3O7/c1-8(20)16-11-13(22)12(21)10(7-19)24-14(11)18-25-15(23)17-9-5-3-2-4-6-9/h2-6,10-13,19,21-22H,7H2,1H3,(H,16,20)(H,17,23)/b18-14-/t10-,11-,12-,13-/m1/s1

InChI key

PBLNJFVQMUMOJY-JXZOILRNSA-N

Looking for similar products? Visit Product Comparison Guide

Application

O-(2-acetamido-2deoxy-D-glucopyranosylidene)amino-N-phenylcarbamate (PUGNAc) has been used to evaluate the effects of silibinin on O-GlcNAc levels of glycoproteins in Adult Retinal Pigment Epithelial-19 (ARPE-19) cells. It has also been used as a component of the HEPES lysis buffer for rat brain samples.
PUGNAc has been used as an inhibitor of O-GlcNAc-β-N-acetylglucosaminidase.

Biochem/physiol Actions

O-(2-acetamido-2deoxy-D-glucopyranosylidene)amino-N-phenylcarbamate (PUGNAc) induces insulin resistance in 3T3-L1 adipocytes by reducing insulin-prompting phosphorylation of protein kinase B (Akt) and glycogen synthase kinase 3β (GSK3β).
PUGNAc is an in vitro and in vivo inhibitor of peptide O-GlcNAc-β-N-acetylglucosaminidase, the enzyme which removes O-linked N-acetylglucosamine (O-GlcNAc) from O-linked glycosylated proteins.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

O-GlcNAc modification on IRS-1 and Akt2 by PUGNAc inhibits their phosphorylation and induces insulin resistance in rat primary adipocytes
Park S Y, et al.
Experimental & Molecular Medicine, 37(3), 220-220 (2005)
Ching-Wen Ho et al.
ACS chemical biology, 5(5), 489-497 (2010-03-02)
Human N-acetyl-beta-hexosaminidase (Hex) isozymes are considered to be important targets for drug discovery. They are directly linked to osteoarthritis because Hex is the predominant glycosidase released by chondrocytes to degrade glycosaminoglycan. Hex is also associated with lysosomal storage disorders. We
Shunichi Yokoe et al.
Glycobiology, 20(10), 1217-1226 (2010-05-21)
Cardiac-type sarco(endo)plasmic reticulum Ca(2)-ATPase (SERCA2a) plays a major role in cardiac muscle contractility. Phospholamban (PLN) regulates the function of SERCA2a via its Ser(16)-phosphorylation. Since it has been proposed that the Ser/Thr residues on cytoplasmic and nuclear proteins are modified by
Matthew S Macauley et al.
Biochimica et biophysica acta, 1800(2), 107-121 (2009-08-12)
The O-GlcNAc modification is found on many nucleocytoplasmic proteins. The dynamic nature of O-GlcNAc, which in some ways is reminiscent of phosphorylation, has enabled investigators to modulate the stoichiometry of O-GlcNAc on proteins in order to study its function. Although
Roselle Gélinas et al.
Nature communications, 9(1), 374-374 (2018-01-27)
AMP-activated protein kinase (AMPK) has been shown to inhibit cardiac hypertrophy. Here, we show that submaximal AMPK activation blocks cardiomyocyte hypertrophy without affecting downstream targets previously suggested to be involved, such as p70 ribosomal S6 protein kinase, calcineurin/nuclear factor of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service