Skip to Content
Merck
All Photos(2)

Key Documents

P5726

Sigma-Aldrich

Phosphatase Inhibitor Cocktail 2

aqueous solution (dark coloration may develop upon storage, which does not affect the activity), for the inhibition of tyrosine protein phosphatases, acid and alkaline phosphatases

Synonym(s):

Protease inhibitor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.54

product name

Phosphatase Inhibitor Cocktail 2, aqueous solution (dark coloration may develop upon storage, which does not affect the activity)

form

aqueous solution (dark coloration may develop upon storage, which does not affect the activity)

Quality Level

shipped in

wet ice

storage temp.

2-8°C

General description

Phosphatase Inhibitor Cocktail 2 is a mixture of components designed to inhibit enzymes that modify proteins present in cell extracts, leading to improved yields of intact proteins. It has been optimized and tested for use on various animal tissues and cytosolic extracts.

Application

The phosphatase inhibitor cocktail improves the yields of intact proteins by adding inhibitors to enzymes that modify proteins present in cell extracts. This product has been optimized and tested for tyrosine protein phosphatases, acid and alkaline phosphatases. It has been tested on cell extracts from animal tissues including bovine liver and human placenta, and cytosolic extract of rabbit muscle.

Biochem/physiol Actions

This mixture contains individual components with specific inhibitory properties. Sodium orthovanadate inhibits a number of ATPases, protein tyrosine phosphatases, and other phosphate-transferring enzymes. Sodium molybdate inhibits acid and phosphoprotein phosphatases. Sodium tartrate inhibits acid phosphatases. Imidazole inhibits alkaline phosphatases.

Features and Benefits

This compound is a featured product for Kinase Phosphatase Biology research. Click here to discover more featured Kinase Phosphatase Biology products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Caution

As supplied, this product is stable for two years at the recommended 2-8°C storage temperature.

Preparation Note

This cocktail is supplied as a clear aqueous solution. It has been sterile filtered through a 0.2 μm membrane in bottles that have been aseptically filled. One mL will inhibit phosphatase activities found in the 100,000 x g supernatant from human placenta, bovine liver, rabbit muscle, A431, or Jurkat cell extracts at a protein concentration of approximately 5 mg/mL.

also commonly purchased with this product

Product No.
Description
Pricing

related product

Product No.
Description
Pricing

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Colin S McCoin et al.
American journal of physiology. Endocrinology and metabolism, 317(2), E298-E311 (2019-05-01)
The impact of sexual dimorphism and mitophagy on hepatic mitochondrial adaptations during the treatment of steatosis with physical activity are largely unknown. Here, we tested if deficiencies in liver-specific peroxisome proliferative activated-receptor-γ coactivator-1α (PGC-1α), a transcriptional coactivator of biogenesis, and
Blanca Majem et al.
Oncogene, 38(32), 6035-6050 (2019-07-07)
Ovarian cancer is the most lethal gynecological malignancy due to the silent nature on its early onset and the rapid acquisition of drug resistance. Histologically heterogeneous, it includes several subtypes with different mutational landscapes, hampering the development of effective targeted
Shira Simonovitch et al.
Journal of Alzheimer's disease : JAD, 70(3), 861-875 (2019-07-16)
This study examined the effects of apolipoprotein E4 (APOE4), the most prevalent genetic risk factor for Alzheimer's disease (AD), on proteins involved in mitochondrial dynamics and autophagy, in the hippocampus of targeted replacement mice. Immunohistochemical measurements revealed that the levels
Uchenna John Unachukwu et al.
Scientific reports, 6, 22392-22392 (2016-03-05)
To replace photoreceptors lost to disease or trauma and restore vision, laboratories around the world are investigating photoreceptor replacement strategies using subretinal transplantation of photoreceptor precursor cells (PPCs) and retinal progenitor cells (RPCs). Significant obstacles to advancement of photoreceptor cell-replacement
Miaozong Wu et al.
PloS one, 4(7), e6430-e6430 (2009-07-31)
Aged skeletal muscle is characterized by an increased incidence of metabolic and functional disorders, which if allowed to proceed unchecked can lead to increased morbidity and mortality. The mechanism(s) underlying the development of these disorders in aging skeletal muscle are

Articles

The use of phosphatase inhibitors is critical for all types of phosphorylation studies. Compare phosphatase inhibitor cocktails to select the best one for your research.

Discover Bioactive Small Molecules for Kinase Phosphatase Biology

Discover Bioactive Small Molecules for Kinase Phosphatase Biology

Discover Bioactive Small Molecules for Kinase Phosphatase Biology

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service