Recommended Products
Related Categories
General description
Alkaline Phosphatase Stabilizing Buffer can extend the shelf life of alkaline phosphate conjugates.
Application
Alkaline Phosphatase (ALP) Stabilizing Buffer may be used in ALP assay in osteoblast cell protein lysates. It may also be used as a component of antibody-enzyme conjugate dialysis buffer.
Alkaline Phosphatase Stabilizing Buffer has been tested in ELISA systems. The product has been specially made for the stable dilution of alkaline phosphatase-antibody conjugates.It can be used for a series of experiments to achieve consistent results.
Components
Stabilizing compounds in Tris buffered saline, pH 8.0, containing 1% bovine serum albumin and 15 mM sodium azide.
Preparation Note
The degree of stability realized is highly dependent upon the conjugate tested. Trials using each particular conjugate should be performed to determine the degree of stability that can be expected. It is recommended that this buffer constitute ≥95% of the final solution.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Storage Class Code
10 - Combustible liquids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Shi Bai et al.
Scientific reports, 8(1), 1048-1048 (2018-01-20)
Guanylate Binding Proteins (GBPs) are a group of cytokine-inducible large guanosine triphosphatase. Previous studies have shown high expression of GBP1 in circulating monocytes of premenopausal subjects was correlated to extremely low peak bone mass, which is considered as an important
Development of biosensor array for rapid detection of cardiac markers: Immunosensor for detection of free cardiac troponin I
ORegan T, et al.
Analytical Letters, 36(9), 1903-1920 (2003)
Xuefeng Zhao et al.
Scientific reports, 6, 19296-19296 (2016-01-15)
Mesenchymal stem cells (MSCs) are multipotent cells, which can give rise to variety of cell types, including adipocytes and osteoblasts. Previously, we have shown that cysteine dioxygenase type 1 (Cdo1) promoted adipogenesis of primary mouse bone marrow stromal cells (BMSCs)
Tanu Sharma et al.
Nutrition and cancer, 72(5), 873-883 (2019-08-15)
Microcalcification seems to be an assurance signature for the prediction of breast cancer malignancy. However, neither systematic study for deciphering the molecular mechanism of mammary microcalcification has yet been conducted, nor a mechanistic study has been performed to find out
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service