Skip to Content
Merck
All Photos(3)

Key Documents

A4675

Sigma-Aldrich

Agar

Type E, suitable for plant cell culture

Synonym(s):

Agar-agar, Gum agar

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C12H18O9)n
CAS Number:
EC Number:
MDL number:
UNSPSC Code:
50405901
eCl@ss:
42040102
NACRES:
NA.21

biological source

algae (Rhodophyceae)

Quality Level

type

Type E

form

powder

technique(s)

cell culture | plant: suitable

transition temp

transition temp 32-38 °C (1.5%)

application(s)

agriculture

storage temp.

room temp

InChI

1S/C14H24O9/c1-5-8(16)13-11(7(21-5)4-20-13)23-14-10(18)12(19-2)9(17)6(3-15)22-14/h5-18H,3-4H2,1-2H3/t5?,6-,7?,8-,9+,10-,11?,12+,13+,14?/m1/s1

InChI key

GYYDPBCUIJTIBM-DYOGSRDZSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Agar or Type E agar is a thermo-reversible, polysaccharide gelling agent. Structurally it consists of two main fractions about 70% of agarose and 30% of agaropectin. The gelling properties of agar comes from the agarose, the high gel strength backbone consists of a linear polymer of 3-linked β-D-galactose and 4-linked 3,6-anhydro-α-L-galactose (LA) repeating units.
Agar or Type E agar is a thermo-reversible, polysaccharide gelling agent. Structurally, it consists of two main fractions about 70% of agarose and 30% of agaropectin. The gelling properties of agar comes from the agarose, the high gel strength backbone consists of a linear polymer of 3-linked ß-D-galactose and 4-linked 3,6-anhydro-α-L-galactose (LA) repeating units.

Application

Agar has been used as reagent for the following studies:
  • Plant cell culture studies.
  • As medium supplement for the growth of wild-type Arabidopsis thaliana (ecotype Columbia) and transgenic seeds.
  • For the solidification of culture medium of cork oak somatic embryos.
  • to examine its effect on metal(loid) sensitivity of the Arabidopsis thaliana metal-hypersensitive mutants
Agar suspension culture may be used for the analysis of cell transformation of palyoma virus. It might have been used for incubation of single antibody-forming cell to investigate the plaque formation.
Agar has been used to examine its effect on metal(loid) sensitivity of the Arabidopsis thaliana metal-hypersensitive mutants.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Melissa S LeBlanc et al.
Journal of biotechnology, 163(1), 1-9 (2012-10-31)
Arsenic is a toxic metalloid and recognized carcinogen. Arsenate and arsenite are the most common arsenic species available for uptake by plants. As an inorganic phosphate (Pi) analog, arsenate is acquired by plant roots through endogenous Pi transport systems. Inside
Pere Puigderrajols et al.
Journal of experimental botany, 53(373), 1445-1452 (2002-05-22)
The timing and tissue localization of small heat shock proteins (sHSPs) during cork oak somatic embryo development was investigated under normal growing culture conditions and in response to stress. Western blot analyses using polyclonal antibodies raised against cork oak recombinant
Human bone marrow colony growth in agar-gel.
B L Pike et al.
Journal of cellular physiology, 76(1), 77-84 (1970-08-01)
AGAR SUSPENSION CULTURE FOR THE SELECTIVE ASSAY OF CELLS TRANSFORMED BY POLYOMA VIRUS.
I MACPHERSON et al.
Virology, 23, 291-294 (1964-06-01)
Selection of Agar Reagents for Medium Solidification Is a Critical Factor for Metal (loid) Sensitivity and Ionomic Profiles of Arabidopsis thaliana
Uraguchi S, et al.
Frontiers in Plant Science, 11, 503-503 (2020)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service