Skip to Content
Merck
All Photos(2)

Key Documents

A2142

Sigma-Aldrich

N-Acetyl-α-D-glucosamine 1-phosphate disodium salt

≥95% (TLC)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C8H14NO9PNa2
CAS Number:
Molecular Weight:
345.15
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

Quality Level

Assay

≥95% (TLC)

form

powder

impurities

≤9% water (Karl Fischer)

color

white

solubility

water: 50 mg/mL, clear, colorless

cation traces

Na: 10.0-14.0% (Dry basis)
P: 8.0-9.6% (dry basis)

storage temp.

−20°C

SMILES string

[Na].CC(=O)NC1C(O)C(O)C(CO)OC1OP(O)(O)=O

InChI

1S/C8H16NO9P.Na.H/c1-3(11)9-5-7(13)6(12)4(2-10)17-8(5)18-19(14,15)16;;/h4-8,10,12-13H,2H2,1H3,(H,9,11)(H2,14,15,16);;

InChI key

VFEAQNOAPUQCLS-UHFFFAOYSA-N

General description

N-Acetyl-D-glucosamine 1-phosphate (GlcNAc-1P) acts as a substrate for UDP-GlcNAc pyrophosphorylase to synthesize UDP-GlcNAc in the presence of UTP. GlcNAc-1P is a product of reverse pyrophosphorolysis of UDP-GlcNAc catalyzed by UDP-GlcNAc pyrophosphorylase.

Application

N-Acetyl-α-D-glucosamine 1-phosphate disodium salt has been used as a component of substrate solution for nucleotide triphosphate (NTP) transferases.

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Measurement of Nucleotide Triphosphate Sugar Transferase Activity via Generation of Pyrophosphate
Maeda K, et al.
2013 Nurse's Drug Handbook, 5(8), e1450-e1450 (2015)
Alexandra Deli et al.
The FEBS journal, 277(13), 2740-2753 (2010-05-25)
The genomes of Bacillus cereus and its closest relative Bacillus anthracis each contain two LmbE protein family homologs: BC1534 (BA1557) and BC3461 (BA3524). Only a few members of this family have been biochemically characterized including N-acetylglucosaminylphosphatidyl inositol (GlcNAc-PI), 1-D-myo-inosityl-2-acetamido-2-deoxy-alpha-D-glucopyranoside (GlcNAc-Ins)
A mechanism-inspired UDP-N-acetylglucosamine pyrophosphorylase inhibitor
Raimi OG, et al.
RSC medicinal chemistry, 1(1), 13-25 (2020)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service