Skip to Content
Merck
All Photos(2)

Key Documents

A1824

Sigma-Aldrich

N-Acetyl-5-hydroxytryptamine

≥99% (HPLC), powder, antioxidant

Synonym(s):

N-Acetylserotonin, Normelatonin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H14N2O2
CAS Number:
Molecular Weight:
218.25
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.32

Product Name

N-Acetyl-5-hydroxytryptamine, ≥99% (HPLC), powder

Quality Level

Assay

≥99% (HPLC)

form

powder

color

white to faint yellow

mp

120-122 °C (lit.)

solubility

ethanol: 50 mg/mL

storage temp.

2-8°C

SMILES string

CC(=O)NCCc1c[nH]c2ccc(O)cc12

InChI

1S/C12H14N2O2/c1-8(15)13-5-4-9-7-14-12-3-2-10(16)6-11(9)12/h2-3,6-7,14,16H,4-5H2,1H3,(H,13,15)

InChI key

MVAWJSIDNICKHF-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

N-Acetylserotonin (NAS)/normelatonin acts as a precursor of melatonin in the tryptophan metabolic pathway.

Application

N-Acetyl-5-hydroxytryptamine has been used to treat neuron cells and to analyze its effects on Krüppel-like factor 15 (KLF15) expression.

Biochem/physiol Actions

N-acetyl-serotonin (NAS/normelatonin) can act as a shelter to neurons due to its protecting ability against oxidative challenges. It can also repress the actions of the transcription factor NF-kappaB. NAS possesses antioxidant and antiaging actions. It has protective action against β-amyloid induced neurotoxicity. It helps to maintain the optimal fluidity of the biological membranes.
Immediate precursor of melatonin. It is formed from serotonin and acetyl-CoA in a reaction catalyzed by serotonin N-acetyl transferase, the rate-limiting enzyme in melatonin biosynthesis. This indoleamine is a weak agonist at melatonin receptors, and has moderate effects on G-protein stimulation and inhibition of cAMP accumulation.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N-acetyl-serotonin (normelatonin) and melatonin protect neurons against oxidative challenges and suppress the activity of the transcription factor NF-kappaB
Lezoualch F, et al.
Journal of Pineal Research, 24(3), 168-178 (1998)
Gianluca Tosini et al.
The Neuroscientist : a review journal bringing neurobiology, neurology and psychiatry, 18(6), 645-653 (2012-05-16)
N-Acetylserotonin (NAS) is a naturally occurring chemical intermediate in biosynthesis of melatonin. Previous studies have shown that NAS has different brain distribution patterns from those of serotonin and melatonin, suggesting that NAS might have functions other than as a precursor
J Vanecek et al.
Endocrinology, 126(3), 1509-1513 (1990-03-01)
Melatonin's transduction mechanisms were investigated using in vitro cultured anterior hemipituitaries. Melatonin inhibited cAMP and 3',5'-cyclic GMP accumulation in neonatal rat anterior pituitary stimulated with LHRH. Maximal inhibitory effect was reached within 25 min and persisted for at least 20
Jimo Borjigin et al.
Molecular and cellular endocrinology, 349(1), 13-19 (2011-07-26)
The pineal gland is a neuroendocrine organ of the brain. Its main task is to synthesize and secrete melatonin, a nocturnal hormone with diverse physiological functions. This review will focus on the central and pineal mechanisms in generation of mammalian
KLF15 regulates dopamine D2 receptor and participates in mouse models of neuropathic pain.
Junfei Z, et al.
Biochemical and Biophysical Research Communications, 492(2), 269-274 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service