Skip to Content
Merck
All Photos(2)

Key Documents

93422

Sigma-Aldrich

Triton X-114

Synonym(s):

(1,1,3,3-Tetramethylbutyl)phenyl-polyethylene glycol, Polyethylene glycol tert-octylphenyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H4O)n C14H22O, n = 7 or 8
CAS Number:
Beilstein:
2315025
MDL number:
UNSPSC Code:
12161900
PubChem Substance ID:
NACRES:
NA.21

description

non-ionic

Quality Level

form

liquid

mol wt

average mol wt 537

impurities

≤0.001% peroxides (as H2O2)
≤1% water

ign. residue

≤0.2%

refractive index

n20/D 1.488 (lit.)
n20/D 1.495

CMC

0.2 mM (20-25°C)

transition temp

pour point ~−9 °C
cloud point 23 °C

density

1.052 g/mL at 20 °C
1.058 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤15 mg/kg
K: ≤500 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Na: ≤500 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg

HLB

12.4

SMILES string

CC(C)(C)CC(C)(C)c1ccc(OCCOCCOCCOCCOCCOCCOCCO)cc1

InChI

1S/C28H50O8/c1-27(2,3)24-28(4,5)25-6-8-26(9-7-25)36-23-22-35-21-20-34-19-18-33-17-16-32-15-14-31-13-12-30-11-10-29/h6-9,29H,10-24H2,1-5H3

InChI key

HNLXNOZHXNSSPN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Triton X-114 is a nonionic detergent. It is a good detergent for solubilizing membranes having reversible phase-separation property useful for the separation of intrinsic membrane proteins. Solution of Triton X-114 is homogeneous at 0°C but phase separation into aqueous phase and a detergent phase occurs above 20°C. Phase separation of proteins in solutions of Triton X-114 has been investigated. This octylphenol ethoxylate is compatible with anionic, cationic, and other nonionic surfactants and chemically stable in most acidic & alkaline solutions.

Application

Triton X-114 has been used:
  • In phase separation of integral membrane proteins
  • In the purification of bacterial inclusion bodies
  • To study the clouding phenomenon in promethazine hydrochloride, an amphiphilic phenothiazine drug
  • To partition conditioned media into insoluble and soluble phases before Western blotting
  • For solubilization of phosphatidylinositol 4-phosphate 5-kinase from bovine brain

Legal Information

Triton is a trademark of The Dow Chemical Company or an affiliated company of Dow

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

483.8 °F - closed cup

Flash Point(C)

251 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Sánchez-Ferrer et al.
Plant physiology, 91(4), 1481-1487 (1989-12-01)
Polyphenoloxidase from grape berries is extracted only by nonionic detergents with a hydrophilic-lipophilic balance between 12.4 and 13.5. The enzyme was partially purified in latent form, free of phenolics and chlorophylls, by using temperature phase partitioning in a solution of
J.G. Pryde
Trends in Biochemical Sciences, 11, 160-160 (1986)
Identification of a new 80 k isoform of phosphatidylinositol 4-phosphate 5-kinase from bovine brain.
Schmidt M, et al.
Febs Letters, 325(3), 201-204 (1993)
M Schmidt et al.
FEBS letters, 325(3), 201-204 (1993-07-05)
Phosphatidylinositol 4-phosphate 5-kinase is associated with bovine brain microsomes to an extent of approximately 65% of the total cellular enzyme activity. This membrane-associated kinase activity can be solubilized with Triton X-114. Following polyacrylamide gel electrophoresis in the presence of SDS
Cloud point variation of amphiphilic drug promethazine hydrochloride with added surfactants.
Naqvi A Z and Khatoon S
Colloids and Surfaces. B, Biointerfaces, 88(2), 568-573 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service