Skip to Content
Merck
All Photos(3)

Key Documents

282103

Sigma-Aldrich

Triton X-100 reduced

Synonym(s):

Polyoxyethylene (10) isooctylcyclohexyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
4-(C8H17)C6H10(OCH2CH2)nOH, n~10
CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

Quality Level

refractive index

n20/D 1.473 (lit.)

density

1.029 g/mL at 25 °C (lit.)

SMILES string

CC(C)(C)CC(C)(C)C1CCC(CC1)OCCOCCOCCOCCOCCOCCOCCO

InChI

1S/C28H56O8/c1-27(2,3)24-28(4,5)25-6-8-26(9-7-25)36-23-22-35-21-20-34-19-18-33-17-16-32-15-14-31-13-12-30-11-10-29/h25-26,29H,6-24H2,1-5H3

InChI key

QQJNBKDKLMCALZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Triton X-100 reduced (RTX-100) is formed when the benzene moiety of TX-100 is fully hydrogenated to a cyclohexane derivative. RTX-100 might enhance enzyme digestion and is also known to exhibit some effects on the photoisomerization of bacteriorhodopsin.
Triton X-100 reduced is a versatile nonionic surfactant with a hydrophilic polyethylene oxide chain, widely utilized as a laboratory detergent. Its applications span various areas, including cell biology and biochemical research. This detergent is commonly employed for tasks such as cell lysis to extract proteins or organelles, permeabilizing the membranes of living cells, and serving as a crucial component in lysis buffers. Triton X-100 plays a key role in the isolation of lipid rafts, contributing to enhanced solubility and dispersibility of substances. With a reduced polyoxyethylene content of approximately 10, Triton X-100 exhibits excellent wetting properties and facilitates emulsification.

In biochemical and cell biology research, Triton X-100 is instrumental in solubilizing membrane-bound proteins and isolating lipid rafts. Its unique properties allow for the preservation of the native conformation of proteins obtained from cellular membranes in solution. Triton X-100 reduced is derived from the full hydrogenation of the benzene moiety of TX-100 to a cyclohexane derivative. This modified version, RTX-100, has demonstrated potential in enhancing enzyme digestion and influencing the photoisomerization of bacteriorhodopsin, showcasing its versatility and utility in advanced research applications.

Application

Triton X-100 reduced has been used:
  • as a component of LB-TT for the extraction of total protein from rat brains
  • in ADP-Glo assay and Cytophos adenosine triphosphatase (ATPase) assay
  • in phosphate-buffered saline (PBS) solution for the permeabilization of fibroblasts in 5′ ethynyl uridine staining, immunofluorescence, and immunolabeling

Features and Benefits

  • Non-ionic surfactant
  • Reduced polyoxyethylene content (~10)
  • Improves solubility and dispersibility of substances
  • Excellent wetting properties
  • Enhances emulsification
  • High purity product for research applications

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Legal Information

Triton is a trademark of The Dow Chemical Company or an affiliated company of Dow

comparable product

Product No.
Description
Pricing

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Electrodilatometry 5: Triton X-100 (reduced form) in cyclohexane
Rappon M and Lin SE
Journal of Molecular Liquids, 116(3), 125-130 (2005)
S J Milder et al.
Biochemistry, 30(7), 1751-1761 (1991-02-19)
Time-resolved difference spectra have been obtained for the photocycle of delipidated bacteriorhodopsin monomers (d-BR) in six different detergent micelle environments that were prepared by two new detergent-exchange techniques. A global kinetic analysis of the photocycle spectra for d-BR in each
J Fernandez et al.
Analytical biochemistry, 218(1), 112-117 (1994-04-01)
An improved and simplified procedure for enzymatic digestion of proteins bound to polyvinylidene difluoride (PVDF) membranes for obtaining internal protein sequence data is presented. This improved procedure is compatible with various enzymes (trypsin, endoproteinase Lys-C, endoproteinase Glu-C, and clostripain) and
Yoshinori Masuo et al.
Methods in molecular biology (Clifton, N.J.), 829, 505-530 (2012-01-11)
Mechanisms underlying behavioral abnormalities of patients with attention-deficit hyperactivity disorder (ADHD) disorder are still unknown. It is worth clarifying alterations in the brain of animal models for ADHD. The animals with neonatal treatment with 6-hydroxydopamine (6-OHDA) and congenic wiggling (Wig)
Yingying Xu et al.
Cell reports, 29(4), 800-809 (2019-10-24)
Cockayne syndrome (CS) is a rare genetic neurodevelopmental disorder, characterized by a deficiency in transcription-coupled subpathway of nucleotide excision DNA repair (TC-NER). Mutation of the Cockayne syndrome B (CSB) gene affects basal transcription, which is considered a major cause of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service