Skip to Content
Merck
All Photos(1)

Key Documents

93337

Sigma-Aldrich

Trizma® acetate

BioUltra, ≥99.0% (NT)

Synonym(s):

TRIS acetate salt, Tris(hydroxymethyl)aminomethane acetate salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2C(CH2OH)3 · CH3COOH
CAS Number:
Molecular Weight:
181.19
Beilstein:
3702918
EC Number:
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NA.25

product line

BioUltra

Quality Level

Assay

≥99.0% (NT)

form

crystalline powder

impurities

insoluble matter, passes filter test

ign. residue

≤0.2%

loss

≤0.2% loss on drying, 20 °C (HV)

pH

6.0-7.0 (25 °C, 0.5 M in H2O)

solubility

H2O: 0.5 M at 20 °C, clear, colorless

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Al: ≤5 mg/kg
As: ≤0.1 mg/kg
Ba: ≤5 mg/kg
Bi: ≤5 mg/kg
Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Li: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Sr: ≤5 mg/kg
Zn: ≤5 mg/kg

λ

0.5 M in H2O

UV absorption

λ: 260 nm Amax: 0.07
λ: 280 nm Amax: 0.05

SMILES string

CC(O)=O.NC(CO)(CO)CO

InChI

1S/C4H11NO3.C2H4O2/c5-4(1-6,2-7)3-8;1-2(3)4/h6-8H,1-3,5H2;1H3,(H,3,4)

InChI key

PIEPQKCYPFFYMG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Trizma is used in the formulation of buffer solutions in the pH range between 7.5 and 8.5. Tris buffer solutions are widely used in cell and molecular biology for processes such as protein and nucleic acid extraction and purification. Trizma based buffers are also in column chromatography and in gel electrophoresis. Trizma acetate is used to make Tris acetic acid buffers that are used as diluents for various assays and as an electrophoresis running buffers.

Other Notes

Recommended buffer for maximum sensitivity of ATP assays with firefly luciferase; Assay of glutamate binding

Legal Information

Trizma is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Monica Cubillos-Rojas et al.
Methods in molecular biology (Clifton, N.J.), 869, 205-213 (2012-05-16)
Polyacrylamide gel electrophoresis (PAGE) is one of the most powerful tools used for protein analysis. We describe the use of Tris-acetate buffer and 3-15% polyacrylamide gradient gels to simultaneously separate proteins in the mass range of 10-500 kDa. We show
Choice of buffer anion for the assay of adenosine 5'-triphosphate using firefly luciferase.
W W Nichols et al.
Analytical biochemistry, 114(2), 396-397 (1981-07-01)
M Ito et al.
Life sciences, 38(12), 1089-1096 (1986-03-24)
[3H]L-glutamic acid binding to microfuge tubes and glass was investigated in four buffers. Background binding to these materials was negligible, but was increased by centrifugation or suction in Tris-HCl and Tris-citrate buffer. This binding was much less or eliminated when
Monica Cubillos-Rojas et al.
Electrophoresis, 31(8), 1318-1321 (2010-03-24)
To separate and analyze giant and small proteins in the same electrophoresis gel, we have used a 3-15% polyacrylamide gradient gel containing 2.6% of the crosslinker bisacrylamide and 0.2 M of Tris-acetate buffer (pH 7.0). Samples were prepared in a
Chi-Lin Li et al.
The Analyst, 137(22), 5222-5228 (2012-10-04)
Oligonucleotide (T30695) modified gold nanoparticles (T30695-Au NPs) have been prepared and employed for quantification of lead ions (Pb(2+)) in blood. The detection of Pb(2+) ions is through the formation of Au-Pb alloys and oligonucleotide-Pb(2+) complexes that catalyze the H(2)O(2)-mediated oxidation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service