Skip to Content
Merck
All Photos(1)

Key Documents

78019

Sigma-Aldrich

L-Phenylalanine

≥99.0% (NT), BioUltra

Synonym(s):

(S)-2-Amino-3-phenylpropionic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
165.19
Beilstein:
1910408
EC Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.26
Pricing and availability is not currently available.

Product Name

L-Phenylalanine, BioUltra, ≥99.0% (NT)

product line

BioUltra

Quality Level

Assay

≥99.0% (NT)

form

powder or crystals

optical activity

[α]20/D −34±0.5°, c = 2% in H2O

impurities

insoluble matter, passes filter test
≤0.3% foreign amino acids

ign. residue

≤0.1% (as SO4)

loss

≤0.1% loss on drying, 110°C

color

white

pH

5.0-7.0 (25 °C, 0.1 M in H2O)

mp

270-275 °C (dec.) (lit.)

solubility

H2O: 0.1 M at 20 °C, clear, colorless

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤200 mg/kg

cation traces

Al: ≤5 mg/kg
As: ≤0.1 mg/kg
Ba: ≤5 mg/kg
Bi: ≤5 mg/kg
Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Li: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
NH4+: ≤100 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Sr: ≤5 mg/kg
Zn: ≤5 mg/kg

absorption

cut-off at 274 nm in H2O at 0.1 M

SMILES string

N[C@@H](Cc1ccccc1)C(O)=O

InChI

1S/C9H11NO2/c10-8(9(11)12)6-7-4-2-1-3-5-7/h1-5,8H,6,10H2,(H,11,12)/t8-/m0/s1

InChI key

COLNVLDHVKWLRT-QMMMGPOBSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Growth requirement of various microorganisms[1].

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

CRC Handbook of Microbiology, 4, 1-1 (1974)
Ann Mullally et al.
Blood, 121(18), 3692-3702 (2013-03-15)
Interferon-α (IFNα) is an effective treatment of patients with myeloproliferative neoplasms (MPNs). In addition to inducing hematological responses in most MPN patients, IFNα reduces the JAK2V617F allelic burden and can render the JAK2V617F mutant clone undetectable in some patients. The
Stefanie Keil et al.
Pediatrics, 131(6), e1881-e1888 (2013-05-22)
Sapropterin dihydrochloride, the synthetic form of 6R-tetrahydrobiopterin (BH4), is an approved drug for the treatment of patients with BH4-responsive phenylketonuria (PKU). The purpose of this study was to assess genotypes and data on the long-term effects of BH4/sapropterin on metabolic
Vimesh A Avlani et al.
American journal of physiology. Endocrinology and metabolism, 304(10), E1097-E1104 (2013-03-28)
In addition to its acute effects on hormone secretion, epithelial transport, and shape change, the calcium-sensing receptor (CaSR) modulates the expression of genes that control cell survival, proliferation, and differentiation as well as the synthesis of peptide hormones and enzymes.
John J Mitchell et al.
Genetics in medicine : official journal of the American College of Medical Genetics, 13(8), 697-707 (2011-05-11)
Phenylalanine hydroxylase deficiency is an autosomal recessive disorder that results in intolerance to the dietary intake of the essential amino acid phenylalanine. It occurs in approximately 1:15,000 individuals. Deficiency of this enzyme produces a spectrum of disorders including classic phenylketonuria

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service