Skip to Content
Merck
All Photos(1)

Key Documents

W358509

Sigma-Aldrich

L-Phenylalanine

99%, FCC

Synonym(s):

(S)-2-Amino-3-phenylpropionic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
165.19
FEMA Number:
3585
Beilstein:
1910408
EC Number:
Council of Europe no.:
10488
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
17.018
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

Kosher

Agency

meets purity specifications of JECFA

reg. compliance

FCC
FDA 21 CFR 172.320

Assay

99%

optical activity

[α]25/D -34.2°, c = 2 in H2O (dried basis)

mp

270-275 °C (dec.) (lit.)

solubility

H2O: soluble 16.5 g/L at 20 °C (completely)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

odorless

SMILES string

N[C@@H](Cc1ccccc1)C(O)=O

InChI

1S/C9H11NO2/c10-8(9(11)12)6-7-4-2-1-3-5-7/h1-5,8H,6,10H2,(H,11,12)/t8-/m0/s1

InChI key

COLNVLDHVKWLRT-QMMMGPOBSA-N

Looking for similar products? Visit Product Comparison Guide

Disclaimer

For R&D or non-EU Food use. Not for retail sale.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vimesh A Avlani et al.
American journal of physiology. Endocrinology and metabolism, 304(10), E1097-E1104 (2013-03-28)
In addition to its acute effects on hormone secretion, epithelial transport, and shape change, the calcium-sensing receptor (CaSR) modulates the expression of genes that control cell survival, proliferation, and differentiation as well as the synthesis of peptide hormones and enzymes.
Ann Mullally et al.
Blood, 121(18), 3692-3702 (2013-03-15)
Interferon-α (IFNα) is an effective treatment of patients with myeloproliferative neoplasms (MPNs). In addition to inducing hematological responses in most MPN patients, IFNα reduces the JAK2V617F allelic burden and can render the JAK2V617F mutant clone undetectable in some patients. The
John J Mitchell et al.
Genetics in medicine : official journal of the American College of Medical Genetics, 13(8), 697-707 (2011-05-11)
Phenylalanine hydroxylase deficiency is an autosomal recessive disorder that results in intolerance to the dietary intake of the essential amino acid phenylalanine. It occurs in approximately 1:15,000 individuals. Deficiency of this enzyme produces a spectrum of disorders including classic phenylketonuria
Stefanie Keil et al.
Pediatrics, 131(6), e1881-e1888 (2013-05-22)
Sapropterin dihydrochloride, the synthetic form of 6R-tetrahydrobiopterin (BH4), is an approved drug for the treatment of patients with BH4-responsive phenylketonuria (PKU). The purpose of this study was to assess genotypes and data on the long-term effects of BH4/sapropterin on metabolic
Bart Pennings et al.
The American journal of clinical nutrition, 98(1), 121-128 (2013-05-03)
Older individuals generally experience a reduced food-chewing efficiency. As a consequence, food texture may represent an important factor that modulates dietary protein digestion and absorption kinetics and the subsequent postprandial protein balance. We assessed the effect of meat texture on

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service