Skip to Content
Merck
All Photos(1)

Key Documents

75044

Sigma-Aldrich

Hyaluronic acid sodium salt from Streptococcus equi

mol wt 150,000-300,000

Synonym(s):

Sodium hyaluronate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.25
Pricing and availability is not currently available.

biological source

(Streptococcus equi)

Quality Level

form

powder

mol wt

150,000-300,000

color

white

suitability

conforms to structure for Infrared spectrum

shipped in

wet ice

storage temp.

−20°C

SMILES string

[Na+].N([C@H]1[C@@H](O[C@@H]([C@H]([C@@H]1O[C@@H]4O[C@@H]([C@H]([C@@H]([C@H]4O)O)O)C(=O)O)O)CO)O[C@@H]2[C@H](O[C@H]([C@@H]([C@H]2O)O)O[C@@H]3[C@H]([C@@H](O[C@@H]([C@H]3O)CO)O)NC(=O)C)C(=O)O)C(=O)C

InChI

1S/C28H44N2O23.Na/c1-5(33)29-9-18(11(35)7(3-31)47-25(9)46)49-28-17(41)15(39)20(22(53-28)24(44)45)51-26-10(30-6(2)34)19(12(36)8(4-32)48-26)50-27-16(40)13(37)14(38)21(52-27)23(42)43;/h7-22,25-28,31-32,35-41,46H,3-4H2,1-2H3,(H,29,33)(H,30,34)(H,42,43)(H,44,45);/q;+1/t7-,8-,9-,10-,11-,12-,13+,14+,15-,16-,17-,18-,19-,20+,21+,22+,25-,26+,27-,28-;/m1./s1

InChI key

YWIVKILSMZOHHF-QJZPQSOGSA-N

Looking for similar products? Visit Product Comparison Guide

Application

suitable as substrate for hyaluronidase

Other Notes

To gain a comprehensive understanding of our extensive range of Polysaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fabrication and Characterization of Vitamin E TPGS Loaded Silk Fibroin/Hyaluronic Acid Nanofibrous Scaffolds.
Ji, L.L., et al.
Advanced Materials Research, 721, 274-277 (2013)
Christine C Krieger et al.
Endocrinology, 155(2), 627-634 (2013-12-05)
Excess production of hyaluronan (hyaluronic acid [HA]) in the retro-orbital space is a major component of Graves' ophthalmopathy, and regulation of HA production by orbital cells is a major research area. In most previous studies, HA was measured by ELISAs
Grit Baier et al.
Biomacromolecules, 14(4), 1103-1112 (2013-03-02)
Antibacterial nanodevices could bring coatings of plastic materials and wound dressings a big step forward if the release of the antibacterial agents could be triggered by the presence of the bacteria themselves. Here, we show that novel hyaluronic acid (HA)-based
Spatially Controlled Delivery of siRNAs to Stem Cells in Implants Generated
Andersen, M.O., et al.
Advances in Functional Materials, 23, 5599-5607 (2013)
Jason A Burdick et al.
Advanced materials (Deerfield Beach, Fla.), 23(12), H41-H56 (2011-03-12)
Hyaluronic acid (HA), an immunoneutral polysaccharide that is ubiquitous in the human body, is crucial for many cellular and tissue functions and has been in clinical use for over thirty years. When chemically modified, HA can be transformed into many

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service