Skip to Content
Merck
All Photos(1)

Key Documents

219991

Sigma-Aldrich

4-Imidazoleacetic acid hydrochloride

98%

Synonym(s):

(4-Imidazolyl)acetic acid hydrochloride, I4AA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H6N2O2 · HCl
CAS Number:
Molecular Weight:
162.57
Beilstein:
3701591
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

mp

218-222 °C (lit.)

solubility

water: soluble 100 mg/mL, clear, faintly yellow

functional group

carboxylic acid

SMILES string

Cl.OC(=O)Cc1c[nH]cn1

InChI

1S/C5H6N2O2.ClH/c8-5(9)1-4-2-6-3-7-4;/h2-3H,1H2,(H,6,7)(H,8,9);1H

InChI key

MWHLCFYPFGFBQO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

4-Imidazoleacetic acid hydrochloride was used in the synthesis of:
  • imidazolyl-polyethylenimine modified nanoparticles
  • pyridyl and imidazoyl functionalized carboproteins, potential metal ion chelators

Biochem/physiol Actions

Competitive antagonist at GABAC receptors.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Archana Swami et al.
International journal of pharmaceutics, 335(1-2), 180-192 (2006-12-16)
The derivatives of polyethylenimine (PEI 25 and 750kDa) were synthesized by partially substituting their amino groups with imidazolyl moieties. The series of imidazolyl-PEIs thus obtained were cross-linked with polyethylene glycol (PEG) to get imidazolyl-PEI-PEG nanoparticles (IPP). The component of hydrophobicity
A Pernille Tofteng et al.
Organic & biomolecular chemistry, 5(14), 2225-2233 (2007-07-05)
De novo design and total chemical synthesis of proteins provides a powerful approach for biological and biophysical studies with the ability to prepare artificial proteins with tailored properties, potentially of importance for biophysical studies, material science, nanobioscience, and as molecular
Karin N Barnouin et al.
Proteomics, 5(17), 4376-4388 (2005-11-19)
IMAC can be used to selectively enrich phosphopeptides from complex peptide mixtures, but co-retention of acidic peptides together with the failure to retain some phosphopeptides restricts the general utility of the method. In this study Fe(III)-IMAC was qualitatively and quantitatively
David I Pattison et al.
Biochemistry, 44(19), 7378-7387 (2005-05-11)
Hypochlorous acid (HOCl) is a powerful oxidant generated from H(2)O(2) and chloride ions by the heme enzyme myeloperoxidase (MPO) released from activated leukocytes. In addition to its potent antibacterial effects, excessive HOCl production can lead to host tissue damage, with
Theoretical studies of IMAC interfacial phenomena for the production of protein C.
E Eileen Thiessen et al.
Advances in experimental medicine and biology, 540, 183-190 (2004-06-04)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service