65984
Micro particles based on polystyrene
size: 800 nm
Synonym(s):
Latex beads from PS
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Quality Level
form
aqueous suspension
concentration
10% (solids)
particle size
800 nm std dev ≤0.03 μm
density
1.05 g/cm3
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
Application
Micro particles based on polystyrene, 800 nM may be derivatized to generate media for applications such as antibody conjugation and use in immunoagglutination assay development or the stabilization of Pickering emulsions. Polystyrene microparticles are useful as potential biodegradable vaccine adjuvants and drug delivery devices.
Analysis Note
For every lot exact values of particle size and standard deviation are determined with an accuracy of 0.01 μm using a Coulter Multisizer.
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Journal of colloid and interface science, 363(1), 307-313 (2011-08-16)
We have studied polydimethylsiloxane (PDMS)-in-1-butyl-3-methylimidazolium hexafluorophosphate ([BMIM][PF(6)]) Pickering emulsions stabilized by polystyrene microparticles with different surface chemistry. Surprisingly, in contrast to the consensus originating from oil/water Pickering emulsions in which the solid particles equilibrate at the oil-water droplet interfaces and
PDA journal of pharmaceutical science and technology, 62(3), 177-190 (2008-07-30)
The objective of this study is to select a multiple-unit sustained-release formulation and to compare it with both commercial immediate and single unit sustained-release capsules and also to determine whether an in vitro-in vivo correlation exists for single- and multiple-
Sensitive Mie scattering immunoagglutination assay of porcine reproductive and respiratory syndrome virus (PRRSV) from lung tissue samples in a microfluidic chip.
J. Virol. Methods, 178, 31-38 (2011)
Proceedings of the National Academy of Sciences of the United States of America, 106(3), 870-875 (2009-01-14)
Many currently used and candidate vaccine adjuvants are particulate in nature, but their mechanism of action is not well understood. Here, we show that particulate adjuvants, including biodegradable poly(lactide-co-glycolide) (PLG) and polystyrene microparticles, dramatically enhance secretion of interleukin-1beta (IL-1beta) by
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service