Skip to Content
Merck
All Photos(1)

Key Documents

60508

Sigma-Aldrich

Potassium disulfite

BioUltra, ≥97.0% (RT)

Synonym(s):

Potassium metabisulfite, Potassium pyrosulfite

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
K2S2O5
CAS Number:
Molecular Weight:
222.32
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.25

product line

BioUltra

Quality Level

Assay

≥97.0% (RT)

impurities

insoluble matter, passes filter test

pH

2.5-4.5 (25 °C, 0.1 M in H2O)

solubility

H2O: 0.1 M at 20 °C, clear, colorless

anion traces

chloride (Cl-): ≤50 mg/kg

cation traces

Al: ≤5 mg/kg
As: ≤0.1 mg/kg
Ba: ≤5 mg/kg
Bi: ≤5 mg/kg
Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
Li: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
Na: ≤1000 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Sr: ≤5 mg/kg
Zn: ≤5 mg/kg

absorption

cut-off at 308 nm in H2O at 0.1 M

SMILES string

[K+].[K+].[O-]S(=O)S([O-])(=O)=O

InChI

1S/2K.H2O5S2/c;;1-6(2)7(3,4)5/h;;(H,1,2)(H,3,4,5)/q2*+1;/p-2

InChI key

RWPGFSMJFRPDDP-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Potassium disulfite (Potassium metabisulfite) may be use to study is antioxidant effects and ability to preserve food. Potassium disulfite may be used as an inhibitor of yeasts, bacteria, and fungi growth. Potassium metabisulfite may be used to study its potential toxicities in vivo.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Supplementary Hazards

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bindu Nair et al.
International journal of toxicology, 22 Suppl 2, 63-88 (2003-10-14)
Sodium Sulfite, Ammonium Sulfite, Sodium Bisulfite, Potassium Bisulfite, Ammonium Bisulfite, Sodium Metabisulfite, and Potassium Metabisulfite are inorganic salts that function as reducing agents in cosmetic formulations. All except Sodium Metabisulfite also function as hair-waving/straightening agents. In addition, Sodium Sulfite, Potassium
D A Mathison et al.
Chest, 87(1 Suppl), 50S-54S (1985-01-01)
Several types of reactions to drugs and chemicals may precipitate or perpetuate asthmatic relapse. This review focuses on reactions to aspirin and sulfites. Approximately 40 percent of patients with rhinosinusitis, nasal polyps, and asthma and 5 to 10 percent of
S Raisuddin et al.
Food additives and contaminants, 8(6), 707-712 (1991-11-01)
The occurrence of aflatoxins in market betel nut samples was studied. It was observed that several betel nut samples were infested with aflatoxin-producing fungus, Aspergillus flavus. Out of 32 samples collected from various places, 12 were positive for aflatoxin. Aflatoxin
Carcinogenicity test of potassium metabisulfite in mice.
T Tanaka et al.
Ecotoxicology and environmental safety, 3(4), 451-453 (1979-12-01)
E Pastorkova et al.
International journal of food microbiology, 161(3), 209-213 (2013-01-22)
This paper investigates the in vitro antimicrobial potential of 15 grape phenolic compounds of various chemical classes (phenolic acids, stilbenes and flavonoids) using the broth microdilution method against yeasts and acetic acid bacteria frequently occurring in deteriorated wine. Pterostilbene (MICs=32-128

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service