Skip to Content
Merck
All Photos(1)

Key Documents

75688

Sigma-Aldrich

Oxalic acid

puriss. p.a., anhydrous, ≥99.0% (RT)

Synonym(s):

Ethanedioic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO2CCO2H
CAS Number:
Molecular Weight:
90.03
Beilstein:
385686
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21

grade

anhydrous
puriss. p.a.

Quality Level

vapor density

4.4 (vs air)

vapor pressure

<0.01 mmHg ( 20 °C)

Assay

≥99.0% (RT)

form

crystals

impurities

≤0.001% total nitrogen (N)
≤1% water

ign. residue

≤0.01% (as SO4)

mp

189.5 °C (dec.) (lit.)

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤350 mg/kg

cation traces

Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤35 mg/kg
K: ≤50 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg

functional group

carboxylic acid

SMILES string

OC(=O)C(O)=O

InChI

1S/C2H2O4/c3-1(4)2(5)6/h(H,3,4)(H,5,6)

InChI key

MUBZPKHOEPUJKR-UHFFFAOYSA-N

Gene Information

human ... SRC(6714)

Looking for similar products? Visit Product Comparison Guide

General description

Oxalic acid is a dicarboxylic acid. It reacts with aqueous iron(II) salt solutions to afford iron(III) oxalate.

Application

Oxalic acid may be used in the preparation of the following:
  • ammonium oxalate
  • calcium oxalate
  • chlorine dioxide (ClO2)
  • potassium hydrogenoxalate

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eagleson M.
Concise Encyclopedia Chemistry, 555-555 (1994)
Eagleson M.
Concise Encyclopedia Chemistry, 66-66 (1994)
Eagleson M.
Concise Encyclopedia Chemistry, 164-164 (1994)
Eagleson M.
Concise Encyclopedia Chemistry, 889-889 (1994)
Eagleson M.
Concise Encyclopedia Chemistry, 215-215 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service