Skip to Content
Merck
All Photos(1)

Key Documents

60279

Sigma-Aldrich

Potassium hexacyanoferrate(II) trihydrate

BioUltra, ≥99.5% (RT)

Synonym(s):

Potassium ferrocyanide, Yellow prussiate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
K4Fe(CN)6 · 3H2O
CAS Number:
Molecular Weight:
422.39
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.25
Assay:
≥99.5% (RT)
form:
solid
solubility:
H2O: 0.5 M at 20 °C, clear, faintly yellow to greenish-yellow

product line

BioUltra

Quality Level

Assay

≥99.5% (RT)

form

solid

impurities

insoluble matter, passes filter test

pH

8.0-10.0 (25 °C, 0.5 M in H2O)

mp

70 °C (lit.)

solubility

H2O: 0.5 M at 20 °C, clear, faintly yellow to greenish-yellow

anion traces

carbonate (CO32-): ≤20 mg/kg
chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

As: ≤0.1 mg/kg
Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤10 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Na: ≤100 mg/kg
Ni: ≤5 mg/kg
Pb: ≤10 mg/kg
Sr: ≤5 mg/kg
Zn: ≤10 mg/kg

absorption

cut-off at 411 nm in H2O at 0.5 M

SMILES string

O.O.O.[K+].[K+].[K+].[K+].N#C[Fe-4](C#N)(C#N)(C#N)(C#N)C#N

InChI

1S/6CN.Fe.4K.3H2O/c6*1-2;;;;;;;;/h;;;;;;;;;;;3*1H2/q;;;;;;-4;4*+1;;;

InChI key

NRYOVBQTVIWDNU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Potassium hexacyanoferrate(II) a redox couple with potassium hexacyanoferrate(III) is used with luminol-based chemiluninescent assays.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jianxiu Du et al.
Luminescence : the journal of biological and chemical luminescence, 17(3), 165-167 (2002-08-08)
A new flow injection chemiluminescence method is described for the determination of captopril. It is based on the enhancing effect of captopril on the chemiluminescence reaction of luminol with potassium ferricyanide in alkaline solution in the presence of potassium ferrocyanide.
Reda R Sheha
Journal of colloid and interface science, 388(1), 21-30 (2012-09-25)
Nanocrystalline potassium zinc hexacyanoferrate loaded on nanoscale magnetite substrate was successfully synthesized for significantly enhanced removal of cesium from low-level radioactive wastes. A description was given for preparation and properties of these precursors. The physicochemical properties of these nanocomposites were
Vasiliy N Goral et al.
Lab on a chip, 6(3), 414-421 (2006-03-03)
A microfluidic biosensor with electrochemical detection for the quantification of nucleic acid sequences was developed. In contrast to most microbiosensors that are based on fluorescence for signal generation, it takes advantage of the simplicity and high sensitivity provided by an
Takeshi Hasegawa et al.
Biomedical chromatography : BMC, 24(6), 613-619 (2009-10-10)
A simple, sensitive and specific chemiluminescent high-performance liquid chromatography method, based on the luminol reaction, for determination of serum cortisol and cortisone, was established. In infants, placental 11beta-hydroxysteroid dehydrogenase type 2 enzyme (11beta-HSD2) activity may affect adrenal function early after
Catarina M Abreu et al.
Biosensors & bioelectronics, 157, 112144-112144 (2020-04-07)
In vitro fertilization (IVF) is the most common assisted reproductive technology used to treat infertility. Embryo selection for transfer in IVF cycles relies on the morphological evaluation by embryologists, either by conventional microscopic assessment or more recently by time-lapse imaging

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service