Skip to Content
Merck
All Photos(3)

Key Documents

P8131

Sigma-Aldrich

Potassium hexacyanoferrate(III)

ReagentPlus®, ~99%

Synonym(s):

Potassium ferricyanide, Red prussiate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
K3Fe(CN)6
CAS Number:
Molecular Weight:
329.24
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
Assay:
~99%
form:
powder or crystals

Quality Level

product line

ReagentPlus®

Assay

~99%

form

powder or crystals

pH

6-9 (25 °C, 329 g/L)

SMILES string

[K+].[K+].[K+].N#C[Fe-3](C#N)(C#N)(C#N)(C#N)C#N

InChI

1S/6CN.Fe.3K/c6*1-2;;;;/q;;;;;;-3;3*+1

InChI key

MIMJFNVDBPUTPB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Potassium hexacyanoferrate(III) can be used as a one-electron oxidant for the oxidation of quinolines, thiamines, and cyclic amines.

Application


  • Enzymatic synthesis of a skin active ingredient - glochidone by 3-ketosteroid dehydrogenase from Sterolibacterium denitrificans: Highlighting the utilization of Potassium hexacyanoferrate(III) in biochemical applications, specifically in enzymatic reactions involving steroid conversion processes (Wojtkiewicz AM et al., 2024).

  • ZIFs-Derived Hollow Nanostructures via a Strong/Weak Coetching Strategy for Long-Life Rechargeable Zn-Air Batteries: Demonstrates the role of Potassium hexacyanoferrate(III) in creating nanostructured materials for enhanced electrochemical energy storage, pertinent to battery technology (Li S et al., 2024).

  • Development of an electrochemical impedance spectroscopy immunosensor for insulin monitoring employing pyrroloquinoline quinone as an ingestible redox probe: Incorporates Potassium hexacyanoferrate(III) for developing advanced biosensors with applications in healthcare diagnostics (Khanwalker M et al., 2024).

  • Selective gold extraction from electronic waste using high-temperature-synthesized reagents: This study uses Potassium hexacyanoferrate(III) in novel methodologies for the recovery of precious metals from electronic waste, offering a sustainable approach to waste management and recycling (Li J et al., 2024).

  • Tunable Electrochemical Entropy through Solvent Ordering by a Supramolecular Host: Explores the chemical versatility of Potassium hexacyanoferrate(III) in creating responsive molecular environments for energy systems, underlining its importance in materials science and thermodynamic studies (Xia KT et al., 2023).

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2

Supplementary Hazards

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Kinetics of Oxidation of Some Fluoroquinolones by Hexacyanoferrate (III) in Alkaline Medium
Diab N, et al.
International journal of chemistry research, 34, 1388-1388 (2013)
Behaviour of cooxidation of isopropyl ether of vitamin A with cumene in chlorobenzene
VaRdanyana R, et al.
Oxidation Communications, 36, 845-851 (2013)
Osmium (VIII)-catalyzed oxidation of some cyclic amines by potassium hexacyanoferrate (III) in alkaline media: A kinetics and mechanistic study
Al-Subu M, et al.
Chemistry of Heterocyclic Compounds, 39, 478-484 (2003)
Simultaneous detection of thiamine and its phosphate esters from microalgae by HPLC
Pinto E, et al.
Biochemical and Biophysical Research Communications, 291, 344-348 (2002)
Willi L Wagner et al.
PloS one, 15(9), e0238798-e0238798 (2020-09-18)
The mesothelium is a dynamic and specialized tissue layer that covers the somatic cavities (pleural, peritoneal, and pericardial) as well as the surface of the visceral organs such as the lung, heart, liver, bowel and tunica vaginalis testis. The potential

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service