Skip to Content
Merck
All Photos(1)

Key Documents

58030

Sigma-Aldrich

Iodonitrotetrazolium chloride

BioReagent, ≥97.0% (calc. on dry substance, NT)

Synonym(s):

2-(4-Iodophenyl)-3-(4-nitrophenyl)-5-phenyl-2H-tetrazolium chloride, p-Iodonitrotetrazolium Violet, INT

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H13ClIN5O2
CAS Number:
Molecular Weight:
505.70
Beilstein:
4093224
EC Number:
MDL number:
UNSPSC Code:
12352102
PubChem Substance ID:
NACRES:
NA.25

product line

BioReagent

Quality Level

Assay

≥97.0% (calc. on dry substance, NT)

mp

240 °C (dec.) (lit.)
~245 °C (dec.)

solubility

methanol: water (1:1): 50 mg/mL, very faintly turbid, very deep yellow (hot)

application(s)

histology

SMILES string

[Cl-].[O-][N+](=O)c1ccc(cc1)-[n+]2nc(nn2-c3ccc(I)cc3)-c4ccccc4

InChI

1S/C19H13IN5O2.ClH/c20-15-6-8-16(9-7-15)23-21-19(14-4-2-1-3-5-14)22-24(23)17-10-12-18(13-11-17)25(26)27;/h1-13H;1H/q+1;/p-1

InChI key

JORABGDXCIBAFL-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Iodonitrotetrazolium (INT) is a tetrazolium dye precursor that forms a purple formazan dye on reduction and has been used in a variety of applications. It is considered to have higher reactivity than some tetrazolium compounds, at least with respect to succinate dehydrogenase, with optimal results obtained using a concentration of 0.8 mM INT. INT is used as an electron acceptor for the colorimetric assays of: lactate dehydrogenase, xanthine dehydrogenase, lactyl-CoA dehydrogenase, succinate dehydrogenase, BBM II ketolisomerase, histidinol dehydrogenase and diverse other hydrolases.
Electron acceptor for the colorimetric assay of various dehydrogenases

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christopher Auger et al.
Electrophoresis, 33(7), 1095-1101 (2012-04-28)
Phosphoenolpyruvate (PEP)-dependent kinases are central to numerous metabolic processes and mediate the production of adenosine triphosphate (ATP) by substrate-level phosphorylation (SLP). While pyruvate kinase (PK, EC: 2.7.1.40), the final enzyme of the glycolytic pathway is critical in the anaerobic synthesis
F Mermillod-Blondin et al.
Water research, 39(9), 1687-1698 (2005-05-19)
Biogeochemical processes mediated by microorganisms in river sediments (hyporheic sediments) play a key role in river metabolism. Because biogeochemical reactions in the hyporheic zone are often limited to the top few decimetres of sediments below the water-sediment interface, slow filtration
T J Mankelow et al.
The Biochemical journal, 358(Pt 2), 315-324 (2001-08-22)
Diethyl pyrocarbonate (DEPC), a histidine-modifying reagent, has been utilized to demonstrate the importance of histidine residues in the functioning of proteins. In previous studies of the NADPH oxidase, histidine residues have been determined to be important in the ability of
Sergio Ravasio et al.
Biochemistry, 41(25), 8120-8133 (2002-06-19)
The properties of the recombinant ferredoxin-dependent glutamate synthase of Synechocystis PCC6803 were determined by means of kinetic and spectroscopic approaches in comparison to those exhibited by the bacterial NADPH-dependent enzyme form. The ferredoxin-dependent enzyme was found to be similar to
Yang Hongwei et al.
Ecotoxicology and environmental safety, 59(2), 249-255 (2004-08-26)
Anaerobic biodegradability of organic compounds can be assessed from the changes in organic compounds, the end products, and the activity of microorganisms. In this study, all three of these variables were considered to integrate assessment of the anaerobic biodegradability of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service