Skip to Content
Merck
All Photos(1)

Key Documents

50246

Sigma-Aldrich

Poloxamer 407, micronized

Synonym(s):

Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol), Lμtrol micro 127

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12161900
NACRES:
NA.25

mol wt

~9840-14600 g/mol

Quality Level

composition

oxyethylene content, 71.5-74.9%

impurities

≤0.25% ethylene glycol and diethylene glycol
≤0.4% total ash
≤0.75% water
≤1 ppm ethylene oxide
≤2 ppm arsenic
≤20 ppm heavy metals
50-125 ppm butylhydroxytoluene

ign. residue

≤0.30%

color

APHA: ≤120

particle size distribution

, coeff var ≤10% (residue, No. 140 USS)
, coeff var ≤50% (residue, No. 270 USS)

pH

5.0-7.5(1:10)
5.0-7.5(1:40)

suitability

complies for Infrared spectrum

InChI

1S/C3H6O.C2H4O/c1-3-2-4-3;1-2-3-1/h3H,2H2,1H3;1-2H2

InChI key

RVGRUAULSDPKGF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

The Poloxamer 407 grades are poloxamers from the extensive line of Pluronic® block copolymers. They are widely used as a thickening agent and gel binder.

Analysis Note

residual solvents (GLC-HS)
1,4-dioxane ≤5 ppm, ethylene glycol ≤620 ppm ( verified on random samples only)

Other Notes

average molecular weight/mass 9840-14600 g/mol; average molecular mass 10000-14600 g/mol

Legal Information

Pluronic is a registered trademark of BASF

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Laurien Buitinga et al.
Clinical and experimental rheumatology, 29(5), 783-789 (2011-10-04)
The Time Trade-Off (TTO) is an instrument used for valuing health-related quality of life. This study evaluated the test-retest reliability of a computer TTO in rheumatoid arthritis (RA), and compared the computer with the interview TTO regarding feasibility and agreement.
Tsz Chung Lai et al.
Colloids and surfaces. B, Biointerfaces, 99, 27-37 (2011-10-18)
Polyplex particles formed with plasmid DNA (pDNA) and Pluronic P85-block-poly{N-[N-(2-aminoethyl)-2-aminoethyl]aspartamide} (P85-b-P[Asp(DET)]) demonstrated highly effective transfection ability compared to PEG-based block cationomer, PEG-b-P[Asp(DET)]. Ternary polyplexes comprising PEG-b-P[Asp(DET)], poly(ethylene oxide)-b-poly(propylene oxide)-b-poly(ethylene oxide)-b-P[Asp(DET)] (P(EPE)-b-P[Asp(DET)]) used as an analog of P85-b-P[Asp(DET)], and pDNA were
Xijun Zhang et al.
Journal of Huazhong University of Science and Technology. Medical sciences = Hua zhong ke ji da xue xue bao. Yi xue Ying De wen ban = Huazhong keji daxue xuebao. Yixue Yingdewen ban, 31(6), 842-845 (2011-12-17)
This study examined the effect of P85 (a pluronic block copolymer) and microbubble (MB) ultrasound contrast agents under ultrasound irradiation on gene transfection and expression. The pEGFP plasmids that can encode enhanced green fluorescent protein (pEGFP) served as a report
Houjian Gong et al.
Langmuir : the ACS journal of surfaces and colloids, 28(38), 13590-13600 (2012-09-08)
The block polyethers PEO-PPO-ph-PPO-PEO (BPE) and PPO-PEO-ph-PEO-PPO (BEP) are synthesized by anionic polymerization using bisphenol A as initiator. Compared with Pluronic P123, the aggregation behaviors of BPE and BEP at an air/water interface are investigated by the surface tension and
Jun Song et al.
Water science and technology : a journal of the International Association on Water Pollution Research, 66(4), 792-798 (2012-07-07)
A novel method for the determination of nickel was established by ultrasonic-assisted cloud point extraction (UA-CPE) prior to flame atomic absorption spectrometry (FAAS) analysis. The nickel reacted with N,N'-bis(salicylidene)-1,2-ethanediamine (BSE) to form hydrophobic chelates, which were extracted into the micelles

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service