Skip to Content
Merck
All Photos(1)

Key Documents

22076

Sigma-Aldrich

(−)-Caryophyllene oxide

≥99.0% (sum of enantiomers, GC)

Synonym(s):

β-Caryophyllene epoxide, (−)-Epoxycaryophyllene, (−)-Epoxydihydrocaryophyllene, (1R,4R,6R,10S)-9-Methylene-4,12,12-trimethyl-5-oxatricyclo[8.2.0.04,6]dodecane, trans-Caryophyllene oxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H24O
CAS Number:
Molecular Weight:
220.35
Beilstein:
148213
EC Number:
MDL number:
UNSPSC Code:
12352212
PubChem Substance ID:
NACRES:
NA.25

Quality Level

Assay

≥99.0% (sum of enantiomers, GC)

form

solid

optical activity

[α]20/D −71±1°, c = 2% in chloroform

mp

61-63 °C
62-63 °C (lit.)

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

2-8°C

SMILES string

[H][C@@]12CCC(=C)[C@@]3([H])CC(C)(C)[C@]3([H])CC[C@@]1(C)O2

InChI

1S/C15H24O/c1-10-5-6-13-15(4,16-13)8-7-12-11(10)9-14(12,2)3/h11-13H,1,5-9H2,2-4H3/t11-,12-,13-,15-/m1/s1

InChI key

NVEQFIOZRFFVFW-RGCMKSIDSA-N

Looking for similar products? Visit Product Comparison Guide

General description

(−)-Caryophyllene oxide or β-caryophyllene oxide (BCPO) is a natural plant compound, bicyclic sesquiterpene, and oxidation derivative of β-caryophyllene (BCP). It has a strong wooden odor. BCPO is commonly found in many food and spice plants and essential oils, such as basil, salvia, and Syzygium cordatum. It is widely used as a cosmetic and food additive.

Application

(−)-Caryophyllene oxide has been used:
  • as an antimicrobial agent to study its effects against Gram-positive, Gram-negative and yeast strains
  • as a commercial standard to perform quantification analysis of constitutes from Marchantia polymorpha extracts using gas chromatography with flame-ionization detection (GC-FID)
  • as a reference standard to analyze the enantiomeric ratios of Achillea ligustica essential oil constituents using gas chromatography-mass spectrometry (GC-MS)

Biochem/physiol Actions

β-caryophyllene oxide is a potent anti-cancer agent which also exerts antioxidant, anti-inflammatory, and antiviral properties. It is a strong inhibitor of drug-metabolizing enzyme cytochromes P4503A (CYP3A) activities both in rat and human hepatic microsomes. BCPO also exerts analgesic, antifungal, antibacterial, and genoprotective activities.

Other Notes

Carefully purified sesquiterpene; Stereoselective rearrangement to an allyl alcohol; Transformation to an amine

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Erich Schmidt et al.
Natural product communications, 5(9), 1365-1368 (2010-10-07)
Commercially available aroma samples were evaluated for their olfactory quality by professional perfumers and tested for their antimicrobial activity. Agar diffusion and agar-dilution were used as test methods and a set of two Gram-positive (Staphylococcus aureus and Enterococcus faecalis) and
H.M.R. Hoffmann et al.
Synlett, 337-337 (1991)
Facundo Romani et al.
Current biology : CB, 30(14), 2815-2828 (2020-06-20)
The origin of a terrestrial flora in the Ordovician required adaptation to novel biotic and abiotic stressors. Oil bodies, a synapomorphy of liverworts, accumulate secondary metabolites, but their function and development are poorly understood. Oil bodies of Marchantia polymorpha develop
Hebaallah Mamdouh Hashiesh et al.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie, 140, 111639-111639 (2021-06-07)
The endocannabinoid system (ECS), a conserved physiological system emerged as a novel pharmacological target for its significant role and potential therapeutic benefits ranging from neurological diseases to cancer. Among both, CB1 and CB2R types, CB2R have received attention for its
Jean-Jacques Filippi et al.
Journal of agricultural and food chemistry, 54(17), 6308-6313 (2006-08-17)
The essential oil of Achillea ligustica from Corsica was investigated by gas chromatography (GC) and gas chromatography-mass spectrometry (GC-MS). A total of 82 compounds representing 94.0% of the oil were tentatively identified. The main constituents were the camphane derivatives, representing

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service