Skip to Content
Merck
All Photos(2)

Key Documents

75047

Supelco

Piperine

analytical standard

Synonym(s):

1-Piperoylpiperidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H19NO3
CAS Number:
Molecular Weight:
285.34
Beilstein:
90741
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥97.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

131-135 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

storage temp.

2-8°C

SMILES string

O=C(\C=C\C=C\c1ccc2OCOc2c1)N3CCCCC3

InChI

1S/C17H19NO3/c19-17(18-10-4-1-5-11-18)7-3-2-6-14-8-9-15-16(12-14)21-13-20-15/h2-3,6-9,12H,1,4-5,10-11,13H2/b6-2+,7-3+

InChI key

MXXWOMGUGJBKIW-YPCIICBESA-N

Looking for similar products? Visit Product Comparison Guide

General description

Piperine is a naturally occurring bioactive alkaloid in pepper, which imparts pungency and bitter flavor to it. It has demonstrated health benefits and exhibits therapeutic properties, which include its role as an antimicrobial, antioxidant, antifungal, immunomodulatory and hepatoprotective agent. The inhibition of drug-metabolizing enzymes in hepatic and pulmonary tissues, and the enhancement of drug bioavailability by this compound are reported. It is also found to play an important role in reducing blood glucose, cholesterol and triglycerides.
Piperine is an amide isolated from the plant, Piper longum. It is a known inhibitor of hepatic and intestinal glucuronidation. Piperine is found to exhibit pharmacological properties like antitumor, antipyretic and anti-inflammatory activities.

Application

Piperine may be used as an analytical reference standard for the quantification of the analyte in:
  • Human plasma using high-performance liquid chromatography coupled to electrospray ionization tandem mass spectrometry (HPLC-ESI-MS/MS).
  • Piper nigrum extract using reversed phase high pressure liquid chromatographic method with UV detection (RP-HPLC-UV).
  • Food products using high-performance liquid chromatography with diode array detection (HPLC-DAD).

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A liquid chromatography method for the simultaneous determination of curcumin and piperine in food products using diode array detection
Nagappan VK , et al.
Asian Journal of Research in Chemistry, 2(2), 115-118 (2009)
Preliminary study on inhibition of genotoxicity by piperine in mice
Selvendiran K, et al.
Fitoterapia, 76(3-4), 296-300 (2005)
Validated HPLC?MS/MS method for simultaneous determination of curcumin and piperine in human plasma
Wang M-X, et al.
Tropical Journal of Pharmaceutical Research, 11(4), 621-629 (2012)
Development and validation of rapid RP-HPLC method for estimation of piperine in Piper nigrum L
Upadhyay V, et al.
International Journal of Herbal Medicine, 1, 6-9 (2013)
Influence of piperine on the pharmacokinetics of curcumin in animals and human volunteers
Shoba G, et al.
Planta Medica, 64(04), 353- 356 (1998)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service