Skip to Content
Merck
All Photos(1)

Key Documents

11161

Sigma-Aldrich

Sodium (R)-β-hydroxyisobutyrate

≥96.0%

Synonym(s):

(R)-β-Hydroxyisobutyric acid sodium salt, (R)-3-Hydroxy-2-methylpropionic acid sodium salt, R-HIBA, R-β-HIBA-Na, Sodium (R)-3-hydroxy-2-methylpropionate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H7NaO3
CAS Number:
Molecular Weight:
126.09
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.25

Quality Level

Assay

≥96.0% (GC)
≥96.0%

optical purity

enantiomeric excess: ≥98.0%

composition

sodium, 17.5-19.0%

storage temp.

2-8°C

SMILES string

[Na+].C[C@H](CO)C([O-])=O

InChI

1S/C4H8O3.Na/c1-3(2-5)4(6)7;/h3,5H,2H2,1H3,(H,6,7);/q;+1/p-1/t3-;/m1./s1

InChI key

RBJZIQZDAZLXEK-AENDTGMFSA-M

Looking for similar products? Visit Product Comparison Guide

Application

3-Hydroxyisobutyrate (HIBA) may be used to study the distribution, characterization and kinetics of enzymes involved in branched chain amino acid metabolism such as 3-hydroxyisobutyrate dehydrogenase (EC: 1.1.1.31) and 3-hydroxyisobutyryl-CoA hydrolase (EC: 3.1.2.4). (R)-β-hydroxyisobutyrate may be used in stereospecific studies.

Biochem/physiol Actions

Increased urinary concentrations of 3-hydroxyisobutyric acid are a biochemical hallmark of metabolic disease 3-hydroxyisobutyric aciduria.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Neuropathology of 3-hydroxyisobutyric aciduria, an autopsy case report.
Xianyuan Song et al.
The Canadian journal of neurological sciences. Le journal canadien des sciences neurologiques, 36(4), 483-486 (2009-08-05)
Jörn Oliver Sass et al.
Journal of inherited metabolic disease, 35(3), 437-442 (2011-08-25)
3-hydroxyisobutyric aciduria is an organic aciduria with a poorly understood biochemical basis. It has previously been assumed that deficiency of 3-hydroxyisobutyrate dehydrogenase (HIBADH) in the valine catabolic pathway is the underlying enzyme defect, but more recent evidence makes it likely
Radovan Murín et al.
Journal of neurochemistry, 105(4), 1176-1186 (2008-02-21)
The branched-chain amino acids (BCAAs)--isoleucine, leucine, and valine--belong to the limited group of substances transported through the blood-brain barrier. One of the functions they are thought to have in brain is to serve as substrates for meeting parenchymal energy demands.
Ference J Loupatty et al.
American journal of human genetics, 80(1), 195-199 (2006-12-13)
Only a single patient with 3-hydroxyisobutyryl-CoA hydrolase deficiency has been described in the literature, and the molecular basis of this inborn error of valine catabolism has remained unknown until now. Here, we present a second patient with 3-hydroxyisobutyryl-CoA hydrolase deficiency
Ronald J A Wanders et al.
Journal of inherited metabolic disease, 35(1), 5-12 (2010-11-26)
Valine is one of the three branched-chain amino acids which undergoes oxidation within mitochondria. In this paper, we describe the current state of knowledge with respect to the enzymology of the valine oxidation pathway and the different disorders affecting oxidation.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service