Skip to Content
Merck
All Photos(1)

Key Documents

54920

Sigma-Aldrich

(R)-3-Hydroxybutyric acid

≥98.0% (T)

Synonym(s):

(3R)-3-Hydroxybutanoic acid, (3R)-Hydroxybutyrate, 3R-Hydroxybutanoic acid, D-β-Hydroxybutyrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H8O3
CAS Number:
Molecular Weight:
104.10
Beilstein:
1720568
EC Number:
MDL number:
UNSPSC Code:
51113400
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥98.0% (T)

form

solid

optical activity

[α]20/D −25±1°, c = 6% in H2O

mp

49-50 °C (lit.)

storage temp.

2-8°C

SMILES string

C[C@@H](O)CC(O)=O

InChI

1S/C4H8O3/c1-3(5)2-4(6)7/h3,5H,2H2,1H3,(H,6,7)/t3-/m1/s1

InChI key

WHBMMWSBFZVSSR-GSVOUGTGSA-N

Looking for similar products? Visit Product Comparison Guide

Application

(R)-3-Hydroxybutyric acid may be used in the preparation of copolymers by reacting with various hydroxyalkanoic acids.1 It may also be used in the preparation of (3R,4R)-4-acetoxy-3-[(R)-1-(formyloxy)ethyl]-2-azetidinone.2

Other Notes

Important chiral starting material; enantioselective reactions at the 2-, 3- and 4-positions via the cyclic acetal with aldehydes; preparation of (R)-β-butyrolactone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D. Seebach et al.
Modern Synthetic Methods, 4, 125-125 (1986)
Journal of the American Chemical Society, 110, 4763-4763 (1988)
A. Griesbech et al.
Helvetica Chimica Acta, 70, 1320-1320 (1987)
J L Hansen et al.
Clinical chemistry, 24(3), 475-479 (1978-03-01)
Methods are described for direct assays of lactate, pyruvate, beta-hydroxybutyrate, and acetoacetate in plasma with the GEMSAEC centrifugal analyzer. The methods for lactate, beta-hydroxybutyrate, and acetoacetate are kinetic and ratiometric, eliminating the need for specimen-blank assays. The pyruvate method is
Jin Hee Kim et al.
Diabetes, obesity & metabolism (2018-11-09)
To investigate sodium-glucose cotransporter 2 inhibitor (SGLT2i)-induced changes in ketogenic enzymes and transporters in normal and diabetic mice models. Normal mice were randomly assigned to receive either vehicle or SGLT2i (25 mg/kg/d by oral gavage) for 7 days. Diabetic mice were treated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service