Skip to Content
Merck
All Photos(1)

Key Documents

08124

Sigma-Aldrich

Isoamylase from Pseudomonas sp.

ammonium sulfate suspension, ≥10,000,000 units/mg protein

Sign Into View Organizational & Contract Pricing


About This Item

Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

form

ammonium sulfate suspension

Quality Level

specific activity

≥10,000,000 units/mg protein

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Isoamylase from Pseudomonas sp. has been used in starch debranching and to measure the chain-length distributions.

Biochem/physiol Actions

Pseudomonas amyloderamosa is considered a popular source of isoamylase. In the presence of a surfactant, this isoamylase produces a series of maltooligosaccharides. Immobilized isoamylase makes it applicable with a polysaccharide matrix composed of agarose, cellulose, and raw corn starch.
Catalyzes the hydrolysis of (1-6)-α-D-glucosidic branch linkages in glycogen, amylopectin and their β-limit dextrins.

Unit Definition

One unit causes an increase in A610 of 0.1 in 1 hour using a homogenized solution of rice starch as a substrate (pH 3.5, 40°C)

Analysis Note

≥10 MU/mg protein

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Digestion properties of corn starch modified by $\alpha$-D-glucan branching enzyme and cyclodextrin glycosyltransferase
Li Y, et al.
Food Hydrocolloids, 89, 534-541 (2019)
Enzymatic conversions of starch
Tomasik P and Horton D
Advances in Carbohydrate Chemistry and Biochemistry, 68, 59-436 (2012)
Rupendra Mukerjea et al.
Carbohydrate research, 345(3), 449-451 (2009-12-26)
Amylose, a putative linear alpha-(1-->4)-glucan and a component of most starches, was isolated from potato, rice, and wheat starches by forming the 1-butanol complex in a solution of the starches. It previously had been found that these amyloses were incompletely
Lucio Gullo et al.
Pancreas, 38(4), 396-400 (2009-03-20)
To determine the type and frequency of pancreatic lesions detected by magnetic resonance cholangiopancreatography (MRCP) in subjects with asymptomatic pancreatic hyperenzymemia and to assess for a possible relationship between these lesions and the hyperenzymemia. From January 2005 to May 2008
Sebastian Streb et al.
The Plant cell, 20(12), 3448-3466 (2008-12-17)
Several studies have suggested that debranching enzymes (DBEs) are involved in the biosynthesis of amylopectin, the major constituent of starch granules. Our systematic analysis of all DBE mutants of Arabidopsis thaliana demonstrates that when any DBE activity remains, starch granules

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service