Skip to Content
Merck
All Photos(5)

Key Documents

A0512

Sigma-Aldrich

Amylose from potato

used as amylase substrate

Synonym(s):

α-Amylose

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H10O5)n
CAS Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

biological source

potato

Quality Level

form

powder

impurities

amylopectin, essentially free
≤10% ethanol
≤2% butanol

color

white to off-white

solubility

0.05 M NaOH: 1 mg/mL, clear to slightly hazy, colorless

storage temp.

room temp

InChI

1S/C18H32O16/c19-1-4-7(22)8(23)12(27)17(31-4)34-15-6(3-21)32-18(13(28)10(15)25)33-14-5(2-20)30-16(29)11(26)9(14)24/h4-29H,1-3H2/t4-,5-,6-,7-,8+,9-,10-,11-,12-,13-,14-,15-,16+,17-,18-/m1/s1

InChI key

FYGDTMLNYKFZSV-PXXRMHSHSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Amylose, a ubiquitous carbohydrate polymer in starch, is made up of a long α-1,4-linked linear chain with rare α-1,6-linked branch points. It is usually seen in the amorphous regions of the granule. It has a left-handed α-helical structure with one turn of the helix consisting of six to eight anhydroglucose units.

Application

Amylose from potatoes has been used as a standard to determine the apparent amylose content (AAC) of rice starch by the iodine-binding colorimetric method.

Biochem/physiol Actions

Amylose is a polyglucose polysaccharide that makes starch(s) together with amylopectin. The glucosidic bonds of amylopectin are α-1,4 linkages. Amylose is used to identify, differentiate and characterize amylase(s). Amylose is used as a thickening and gelling agent, and emulsion stabilizer. Amylose is used to prepare films and membranes with potential use in drug delivery.

Substrates

A substrate for serum amylase.

Other Notes

To gain a comprehensive understanding of our extensive range of Polysaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shujun Wang et al.
Scientific reports, 6, 28271-28271 (2016-06-21)
A thorough understanding of starch gelatinization is extremely important for precise control of starch functional properties for food processing and human nutrition. Here we reveal the molecular mechanism of starch gelatinization by differential scanning calorimetry (DSC) in conjunction with a
Caili Li et al.
Carbohydrate polymers, 245, 116557-116557 (2020-07-29)
High-amylose wheat starch (HAWS) and flour (HAWF) have the potential to deliver food products with enhanced nutritional functionality, but structure/function relationships are not well understood. We report the structural bases for differences in water absorption and pasting properties for HAWS
Xingli Liu et al.
Carbohydrate polymers, 283, 119148-119148 (2022-02-15)
In order to provide a theoretical basis on how germination treatment modulated the pasting behaviors of starches from native and germinated waxy brown rice, impacts of germination on the molecular and supramolecular structures and pasting behaviors were evaluated. Multiple analytical
Rachmawati Rachmawati et al.
Biomacromolecules, 14(2), 575-583 (2013-01-16)
Several methods were used to investigate the possibility of preparing inclusion complexes between amylose and polytetrahydrofurans (PTHF) via direct mixing. Potato amylose (M(v) ∼ 200 kg/mol) and synthetic amylose (M(n) 42 kg/mol) were complexed with PTHF having different molecular weights
Ahmed A Younes et al.
Journal of pharmaceutical and biomedical analysis, 75, 74-85 (2013-01-15)
Nine polysaccharide-based chiral stationary phases (CSPs) were used to define a separation strategy that combines normal-phase (NP), reversed-phase (RP) and polar organic solvent chromatography (POSC) modes. After limiting ourselves to two CSPs per mode, in total, five CSPs, Chiralpak AD

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service